Skip to Content
Merck
All Photos(3)

Key Documents

W266523

Sigma-Aldrich

L-Menthol

greener alternative

natural, ≥99%, FCC, FG

Synonym(s):

(1R,2S,5R)-(−)-Menthol, (−)-Menthol, (1R,2S,5R)-2-Isopropyl-5-methylcyclohexanol, 5-Methyl-2-(1-methylethyl)cyclohexanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H20O
CAS Number:
Molecular Weight:
156.27
FEMA Number:
2665
Beilstein:
1902293
EC Number:
Council of Europe no.:
65c
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
2.218
NACRES:
NA.21
Organoleptic:
minty; cooling; peppermint
grade:
FG
Halal
Kosher
natural
food allergen:
no known allergens
Pricing and availability is not currently available.

grade

FG
Halal
Kosher
natural

Quality Level

reg. compliance

EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117

vapor pressure

0.8 mmHg ( 20 °C)

Assay

≥99%

optical activity

[α]20/D −49°, c = 10 in 95% ethanol

greener alternative product characteristics

Less Hazardous Chemical Syntheses
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

bp

212 °C (lit.)

mp

41-45 °C (lit.)

density

0.89 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

greener alternative category

Organoleptic

minty; cooling; peppermint

SMILES string

CC(C)[C@@H]1CC[C@@H](C)C[C@H]1O

InChI

1S/C10H20O/c1-7(2)9-5-4-8(3)6-10(9)11/h7-11H,4-6H2,1-3H3/t8-,9+,10-/m1/s1

InChI key

NOOLISFMXDJSKH-KXUCPTDWSA-N

Looking for similar products? Visit Product Comparison Guide

General description

L-Menthol is the principal component of peppermint oil. It is widely used in confectionaries cosmetics and toothpaste for its characteristic peppermint flavor and cooling effect.[1]

Application

L-Menthol can be used as a model flavor compound to prepare flavor encapsulated porous cellulose particles for food processing applications.[2]

Biochem/physiol Actions

Taste at 25 ppm

Other Notes

Natural occurrence: Mentha species.

Disclaimer

The substance W266523 has no REACH registration because the only supported use is the use in medicinal products for human or veterinary use or in food or feedingstuffs according to article 2 of the REACH Regulation (EC) No 1907/2006.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

201.2 °F - closed cup

Flash Point(C)

94 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

An investigation of flavor encapsulation comprising of regenerated cellulose as core and carboxymethyl cellulose as wall
Ma M, et al.
Iranian Polymer Journal, 22(9), 689-695 (2013)
Optimized synthesis of lipase-catalyzed l-menthyl butyrate by Candia rugosa lipase.
Shih L, et al.
Food Chemistry, 100(3), 1223-1228 (2007)
Thomas Voets et al.
Nature chemical biology, 3(3), 174-182 (2007-02-13)
TRPM8, a member of the transient receptor potential (TRP) channel superfamily, is expressed in thermosensitive neurons, in which it functions as a cold and menthol sensor. TRPM8 and most other temperature-sensitive TRP channels (thermoTRPs) are voltage gated; temperature and ligands
A Gelal et al.
Clinical pharmacology and therapeutics, 66(2), 128-135 (1999-08-25)
Menthol is widely used in a variety of commercial products and foods, but its clinical pharmacology is not well studied. To determine the disposition kinetics and to examine subjective and cardiovascular effects of menthol, we conducted a crossover placebo-controlled study
Tibor Rohács et al.
Nature neuroscience, 8(5), 626-634 (2005-04-27)
The subjective feeling of cold is mediated by the activation of TRPM8 channels in thermoreceptive neurons by cold or by cooling agents such as menthol. Here, we demonstrate a central role for phosphatidylinositol 4,5-bisphosphate (PI(4,5)P(2)) in the activation of recombinant

Questions

  1. What does SAMPLE-K mean?

    1 answer
    1. The Sample-K SKU designation refers to a sample amount of the item. This smaller quantity is often utilized to qualify the material for use without purchasing the larger quantities listed - in this case the 1KG and the 4KG.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service