Skip to Content
Merck
All Photos(3)

Key Documents

W276413

Sigma-Aldrich

Myristic acid

greener alternative

natural, ≥98.5%, FG

Synonym(s):

1-Tridecanecarboxylic acid, C14:0, NSC 5028, Tetradecanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)12COOH
CAS Number:
Molecular Weight:
228.37
FEMA Number:
2764
Beilstein:
508624
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
8.016
NACRES:
NA.21

grade

FG
Fragrance grade
Halal
Kosher
natural

Quality Level

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002

Assay

≥98.5%

greener alternative product characteristics

Less Hazardous Chemical Syntheses
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

bp

250 °C/100 mmHg (lit.)

mp

52-54 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

greener alternative category

Organoleptic

oily; waxy; soapy

SMILES string

CCCCCCCCCCCCCC(O)=O

InChI

1S/C14H28O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14(15)16/h2-13H2,1H3,(H,15,16)

InChI key

TUNFSRHWOTWDNC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Experiments and Molecular Simulations to Study the Effect of Surface-Active Compounds in Mixtures of Model Oils on CO2 Corrosion during Intermittent Oil-Water Wetting.: Investigates how myristic acid, as a surface-active compound, influences CO2 corrosion mechanisms in oil-water systems, offering insights into corrosion prevention strategies in the oil industry (Norooziasl et al., 2024).


Disclaimer

The substance W276413 has no REACH registration because the only supported use is the use in medicinal products for human or veterinary use or in food or feedingstuffs according to article 2 of the REACH Regulation (EC) No 1907/2006.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dalit Shental-Bechor et al.
Proceedings of the National Academy of Sciences of the United States of America, 109(44), 17839-17844 (2012-08-01)
We present an integrated experimental and computational study of the molecular mechanisms by which myristoylation affects protein folding and function, which has been little characterized to date. Myristoylation, the covalent linkage of a hydrophobic C14 fatty acyl chain to the
S Curry et al.
Biochimica et biophysica acta, 1441(2-3), 131-140 (1999-11-26)
Human serum albumin possesses multiple fatty acid binding sites of varying affinities, but the precise locations of these sites have remained elusive. The determination of the crystal structure of human serum albumin complexed with myristic acid recently revealed the positions
Anja Meier et al.
Hepatology (Baltimore, Md.), 58(1), 31-42 (2012-12-06)
Chronic infection with the human hepatitis B virus (HBV) is a global health problem and a main cause of progressive liver diseases. HBV exhibits a narrow host range, replicating primarily in hepatocytes. Both host and hepatocyte specificity presumably involve specific
Sarah Brice Russo et al.
The Journal of clinical investigation, 122(11), 3919-3930 (2012-10-02)
Diabetic cardiomyopathy (DbCM), which consists of cardiac hypertrophy and failure in the absence of traditional risk factors, is a major contributor to increased heart failure risk in type 2 diabetes patients. In rodent models of DbCM, cardiac hypertrophy and dysfunction
Toko Chida et al.
The Biochemical journal, 449(3), 741-749 (2012-10-24)
PPM [metal-dependent protein phosphatase, formerly called PP2C (protein phosphatase 2C)] family members play essential roles in regulating a variety of signalling pathways. While searching for protein phosphatase(s) that act on AMPK (AMP-activated protein kinase), we found that PPM1A and PPM1B

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service