Skip to Content
Merck
All Photos(3)

Key Documents

W200336

Sigma-Aldrich

Acetaldehyde

greener alternative

natural, FG

Synonym(s):

Ethanal

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CHO
CAS Number:
Molecular Weight:
44.05
FEMA Number:
2003
Beilstein:
505984
EC Number:
Council of Europe no.:
89
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
5.001
NACRES:
NA.21
grade:
FG
Halal
Kosher
natural
food allergen:
no known allergens

grade

FG
Halal
Kosher
natural

Quality Level

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117

vapor density

1.52 (vs air)

vapor pressure

14.63 psi ( 20 °C)

Assay

≥99% (GC)

form

liquid

autoignition temp.

365 °F

expl. lim.

60 %

greener alternative product characteristics

Less Hazardous Chemical Syntheses
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

refractive index

n20/D 1.332 (lit.)

pH

5 (20 °C)

bp

21 °C (lit.)

mp

−125 °C (lit.)

density

0.785 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

greener alternative category

Organoleptic

ethereal

storage temp.

2-8°C

SMILES string

CC=O

InChI

1S/C2H4O/c1-2-3/h2H,1H3

InChI key

IKHGUXGNUITLKF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Acetaldehyde is an important volatile flavoring compound found in Sherry-like wines and also in many fruits. It is mainly used as a flavoring ingredient in milk products, fruit juices and soft drinks.
We are committed to bringing you Greener Alternative Products, which adhere to one of the four categories of Greener Alternatives . This product is a Biobased products, showing key improvements in Green Chemistry Principles “Less Hazardous Chemical Syntheses” and “Use of Renewable Feedstock”.

Application


  • Identification of acetaldehyde based on plasmonic patterns of a gold nanostructure conjugated with chromophore and H(2)O(2): a new platform for the rapid and low-cost analysis of carcinogenic agents by colorimetric affordable test strip (CATS).: This study presents a novel method for detecting acetaldehyde using gold nanostructures, offering a fast, affordable solution for monitoring this carcinogenic compound (Shadjou N et al., 2024).

  • Optimisation and characterisation of KOH-activated carbon obtained from Baijiu spent grains for the mitigation of risk factors in alcoholic beverages.: This paper explores the use of KOH-activated carbon for absorbing acetaldehyde from alcoholic beverages, reducing its carcinogenic risk (Sun B et al., 2024).

  • Single-site iron-anchored amyloid hydrogels as catalytic platforms for alcohol detoxification.: Discusses the development of novel iron-anchored amyloid hydrogels that catalyze the breakdown of acetaldehyde, a harmful metabolite of alcohol (Mezzenga R et al., 2024).

Caution

May form a small amount of trimer

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Eye Irrit. 2 - Flam. Liq. 1 - Muta. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-38.0 °F - closed cup

Flash Point(C)

-38.89 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jelen H
Food Flavors: Chemical, Sensory and Technological Properties, 291-291 (2011)
Burdock GA
Encyclopedia of Food and Color Additives, 1, 9-9 (1997)
Yannai S
Dictionary of Food Compounds with CD-ROM: Additives, Flavors, and Ingredients, 7-7 (2012)
Tetsuji Yokoyama et al.
Alcoholism, clinical and experimental research, 29(4), 622-630 (2005-04-19)
Elevated mean corpuscular volume (MCV) is a traditional biological marker for alcohol abuse and alcoholism, but the underlying mechanism is unclear. Three recent epidemiologic studies consistently showed that MCV was elevated by alcohol drinking more markedly among individuals with genetically
Mikko Salaspuro
Journal of digestive diseases, 12(2), 51-59 (2011-03-16)
Aldehyde dehydrogenase (ALDH2) and alcohol dehydrogenase (ADH) gene polymorphisms associating with enhanced acetaldehyde exposure and markedly increased cancer risk in alcohol drinkers provide undisputable evidence for acetaldehyde being a local carcinogen not only in esophageal but also in gastric cancer.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service