Skip to Content
Merck
All Photos(1)

Key Documents

70082

Sigma-Aldrich

Myristic acid

≥98.0% (GC)

Synonym(s):

1-Tridecanecarboxylic acid, C14:0, NSC 5028, Tetradecanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)12COOH
CAS Number:
Molecular Weight:
228.37
Beilstein:
508624
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25
Pricing and availability is not currently available.

Quality Level

Assay

≥98.0% (GC)

form

powder

bp

250 °C/100 mmHg (lit.)

mp

52-54 °C (lit.)
53-56 °C

functional group

carboxylic acid

lipid type

saturated FAs

shipped in

ambient

storage temp.

room temp

SMILES string

CCCCCCCCCCCCCC(O)=O

InChI

1S/C14H28O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14(15)16/h2-13H2,1H3,(H,15,16)

InChI key

TUNFSRHWOTWDNC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

<ul>
<li><strong>Safety assessment of myristic acid as a food ingredient.</strong> Myristic acid was evaluated for its safety as a food ingredient, demonstrating its compliance with health and safety standards and supporting its use in food manufacturing (Burdock and Carabin, 2007).</li>
<li><strong>Example of fatty acid-loaded lipoplex in enhancing in vitro gene transfer efficacies of cationic amphiphile.</strong> This research outlines how myristic acid can be used to improve the efficiency of gene delivery systems in medical research, suggesting a pivotal role in enhancing transfection efficiencies (Majeti et al., 2005).</li>
</ul>

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dale D O Martin et al.
Biochimie, 93(1), 18-31 (2010-11-09)
Myristoylation corresponds to the irreversible covalent linkage of the 14-carbon saturated fatty acid, myristic acid, to the N-terminal glycine of many eukaryotic and viral proteins. It is catalyzed by N-myristoyltransferase. Typically, the myristate moiety participates in protein subcellular localization by
Yanhong Hao et al.
iScience, 24(9), 102974-102974 (2021-08-17)
Asymptomatic infection is a big challenge in curbing the spread of COVID-19. However, its identification and pathogenesis elucidation remain issues. Here, by performing comprehensive lipidomic characterization of serum samples from 89 asymptomatic COVID-19 patients and 178 healthy controls, we screened
Katarzyna Reczyńska et al.
Lipids, 55(2), 117-126 (2020-01-24)
The impact of saturated fatty acids (FA) on viability and properties of malignant and nonmalignant cells has not been studied in detail so far. The present study was aimed at evaluation of the influence of saturated FA (10:0-18:0) on malignant
Marc Heitz et al.
Bioconjugate chemistry, 30(8), 2165-2182 (2019-08-10)
Transfecting nucleic acids into cells is an essential procedure in biological research usually performed using nonviral transfection reagents. Unfortunately, most transfection reagents have polymeric or undisclosed structures and require nonstandard synthetic procedures. Herein we report peptide dendrimers accessible as pure
S Curry et al.
Biochimica et biophysica acta, 1441(2-3), 131-140 (1999-11-26)
Human serum albumin possesses multiple fatty acid binding sites of varying affinities, but the precise locations of these sites have remained elusive. The determination of the crystal structure of human serum albumin complexed with myristic acid recently revealed the positions

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service