Skip to Content
Merck
All Photos(3)

Key Documents

W243914

Sigma-Aldrich

Ethyl hexanoate

greener alternative

natural, ≥98%, FCC, FG

Synonym(s):

Caproic acid ethyl ester, Ethyl caproate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4COOC2H5
CAS Number:
Molecular Weight:
144.21
FEMA Number:
2439
Beilstein:
1701293
EC Number:
Council of Europe no.:
310
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.060
NACRES:
NA.21
grade:
FG
Halal
Kosher
natural
food allergen:
no known allergens
Pricing and availability is not currently available.

grade

FG
Halal
Kosher
natural

Quality Level

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117

vapor density

5 (vs air)

Assay

≥98%

greener alternative product characteristics

Less Hazardous Chemical Syntheses
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

refractive index

n20/D 1.407 (lit.)

bp

168 °C (lit.)

density

0.869 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

greener alternative category

Organoleptic

banana; green; fruity; waxy; pineapple; sweet

SMILES string

CCCCCC(=O)OCC

InChI

1S/C8H16O2/c1-3-5-6-7-8(9)10-4-2/h3-7H2,1-2H3

InChI key

SHZIWNPUGXLXDT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one of the four categories of Greener Alternatives . This product is a Biobased products, showing key improvements in Green Chemistry Principles “Less Hazardous Chemical Syntheses” and “Use of Renewable Feedstock”.

Application


  • Formation of Key Aroma Compounds During 30 Weeks of Ripening in Gouda-Type Cheese Produced from Pasteurized and Raw Milk.: The study investigates the formation of key aroma compounds, including ethyl hexanoate, during the ripening of Gouda-type cheese. The results provide insights into the ripening process and its impact on cheese flavor development (Duensing et al., 2024).

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

127.4 °F - closed cup

Flash Point(C)

53 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Clémentine Lauverjat et al.
Journal of agricultural and food chemistry, 57(21), 9878-9887 (2009-10-13)
Physicochemical properties (partition and diffusion coefficients) involved in the mobility and release of salt and aroma compounds in model cheeses were determined in this study. The values of NaCl water/product partition coefficients highlighted interactions between proteins and NaCl. However, these
Da-Mi Jung et al.
Journal of agricultural and food chemistry, 51(1), 200-205 (2002-12-28)
Changes in the volatility of selected flavor compounds in the presence of nonvolatile food matrix components were studied using headspace solid-phase microextraction (HS-SPME) combined with GC-MS quantification. Time-dependent adsorption profiles to the SPME fiber and the partition coefficients between different
John C Beaulieu et al.
Journal of agricultural and food chemistry, 55(23), 9503-9513 (2007-10-20)
Changes in post-cutting volatiles, quality, and sensory attributes during fresh-cut storage (4 degrees C) of cantaloupe (Cucumis melo L. var. Reticulatus, Naudin, cv. 'Sol Real') harvested at four distinct maturities (1/4-, 1/2-, 3/4-, and full-slip) were investigated after 0, 2
Dong Wang et al.
Bioprocess and biosystems engineering, 30(3), 147-155 (2007-01-26)
The cell-bound lipase from Rhizopus chinensis CCTCC M201021 with high catalysis ability for ester synthesis was located as a membrane-bound lipase by the treatments of Yatalase firstly. In order to improve its synthetic activity in non-aqueous phase, the pretreatments of
Kazuo Aritomi et al.
Bioscience, biotechnology, and biochemistry, 68(1), 206-214 (2004-01-28)
Point mutation of Gly1250Ser (1250S) of the yeast fatty acid synthase gene FAS2 confers cerulenin resistance. This mutation also results in a higher production of the apple-like flavor component ethyl caproate in Japanese sake. We mutated the 1250th codon by

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service