Skip to Content
Merck
All Photos(1)

Key Documents

W280615

Sigma-Aldrich

Octyl acetate

natural, ≥98%, FCC, FG

Synonym(s):

Acetic acid octyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO2(CH2)7CH3
CAS Number:
Molecular Weight:
172.26
FEMA Number:
2806
EC Number:
Council of Europe no.:
197c
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.007
NACRES:
NA.21
grade:
FG
Fragrance grade
natural
food allergen:
no known allergens

grade

FG
Fragrance grade
natural

Quality Level

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 172.515

vapor density

5.9 (vs air)

Assay

≥98%

autoignition temp.

515 °F

expl. lim.

8.14 %

refractive index

n20/D 1.418 (lit.)

bp

211 °C (lit.)

density

0.867 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

floral; fruity; sweet

SMILES string

CCCCCCCCOC(C)=O

InChI

1S/C10H20O2/c1-3-4-5-6-7-8-9-12-10(2)11/h3-9H2,1-2H3

InChI key

YLYBTZIQSIBWLI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Octyl acetate is a compound found in oranges, grapefruits, and other citrus products. Octyl acetate is used for artificial flavors and in perfumery due to its fruity odor.

Application

Octyl acetate is an ester with a fruity orange flavor employed as a flavoring agent in the food and beverage industries.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

189.1 °F - closed cup

Flash Point(C)

87.3 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Esters, terpenes and flavours: Make the mood cheers by three musketeers
Sen DJ
World Journal of Pharmaceutical Research, 4, 01-40 (2015)
Solvent-free synthesis of flavour esters through immobilized lipase mediated transesterification
Garlapati VK, et al.
Enzyme Research, 2013 (2013)
W C Daughtrey et al.
Fundamental and applied toxicology : official journal of the Society of Toxicology, 12(2), 313-320 (1989-02-01)
The subchronic toxicity of octyl acetate was assessed following its administration to rats via oral gavage, 5 days per week for 13 weeks. Treated rats received undiluted octyl acetate at doses of 0.1, 0.5, or 1.0 g/kg. Control rats received
C Ursin et al.
American Industrial Hygiene Association journal, 56(7), 651-660 (1995-07-01)
A procedure has been developed for measuring the steady state rate of permeation of commercial solvents through living human skin. To get the most consistent results, it was necessary with some solvents to normalize the solvent permeation rate of a
Anett Kirschner et al.
Applied microbiology and biotechnology, 73(5), 1065-1072 (2006-09-01)
A gene encoding a Baeyer-Villiger monooxygenase (BVMO) identified in Pseudomonas fluorescens DSM 50106 was cloned and functionally expressed in Escherichia coli JM109. Sodium dodecyl sulfate-polyacrylamide gel electrophoresis (SDS-PAGE) and Western blot analysis showed an estimated 56 kDa-size protein band corresponding

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service