Skip to Content
Merck
All Photos(3)

Key Documents

W269816

Sigma-Aldrich

Methyl cinnamate

greener alternative

natural, ≥98%, FCC, FG

Synonym(s):

Cinnamic acid methyl ester, Methyl 3-phenyl acrylate, Methyl 3-phenylprop-2-enoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH=CHCOOCH3
CAS Number:
Molecular Weight:
162.19
FEMA Number:
2698
Beilstein:
386468
EC Number:
Council of Europe no.:
333c
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.740
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
Kosher
natural
food allergen:
no known allergens

grade

FG
Fragrance grade
Halal
Kosher
natural

Quality Level

300
400

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 172.515

Assay

≥98%

greener alternative product characteristics

Less Hazardous Chemical Syntheses
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

bp

260-262 °C (lit.)

mp

33-38 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

greener alternative category

Organoleptic

balsam; fruity; strawberry

SMILES string

COC(=O)\C=C\c1ccccc1

InChI

1S/C10H10O2/c1-12-10(11)8-7-9-5-3-2-4-6-9/h2-8H,1H3/b8-7+

InChI key

CCRCUPLGCSFEDV-BQYQJAHWSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methyl cinnamate is an important flavoring agent and fragrance ingredient. It is one of the main aroma components of basil oil, Japanese and Korean matsutake mushrooms.
We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product is Biobased and thus aligns with "Less Hazardous Chemical Syntheses" and "Use of Renewable Feedstocks".

Application


  • Methyl Cinnamate (MC) Alleviates Free Fatty Acids (FFAs) Induced Lipid Accumulation Through the AMPK Pathway in HepG2 Cells.: This research demonstrates that Methyl Cinnamate can reduce lipid accumulation in HepG2 cells by activating the AMPK pathway, suggesting potential therapeutic applications for metabolic disorders (Fu et al., 2024).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fragrance material review on methyl cinnamate.
Bhatia SP, et al.
Food And Chemical Toxicology, 45(1), S113-S119 (2007)
Determination of 2H/1H and 13C/12C isotope ratios of (E)-methyl cinnamate from different sources using isotope ratio mass spectrometry.
Fink K, et al.
Agricultural and Biological Chemistry, 52(10), 3065-3068 (2004)
Flavor components in Japanese and Korean matsutake-Tricholoma matsutake (S. Ito et Imai) Sing.-and their change during storage.
Takama F, et al.
Nippon Shokuhin Kogyo Gakkaishi, 31(1), 14-18 (1984)
Basil: a source of essential oils.
Advances in new crops, 484-489 (1990)
Difference in the volatile composition of pine-mushrooms (Tricholoma matsutake Sing.) according to their grades.
Cho IH, et al.
Journal of Agricultural and Food Chemistry, 54(13), 4820-4825 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service