The biological source of this product is confirmed to be botanical. However, it is important to note that there is no C14 report available to certify its bio-carbon content.
W200712
Triacetin
≥99%, natural (US), FG
Synonym(s):
1,2,3-Triacetoxypropane, 1,2,3-Triacetylglycerol, Glyceryl triacetate
About This Item
Fragrance grade
Halal
Kosher
natural (US)
Recommended Products
biological source
botanical
Quality Level
grade
FG
Fragrance grade
Halal
Kosher
natural (US)
Agency
follows IFRA guidelines
reg. compliance
EU Regulation 1223/2009
EU Regulation 1333/2008 & 178/2002
FDA 21 CFR 117
vapor density
7.52 (vs air)
Assay
≥99%
autoignition temp.
809 °F
expl. lim.
1.05 %, 189 °F
7.73 %, 215 °F
greener alternative product characteristics
Less Hazardous Chemical Syntheses
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
refractive index
n25/D 1.429-1.431 (lit.)
bp
258 °C (lit.)
mp
3 °C (lit.)
density
1.16 g/mL at 25 °C (lit.)
application(s)
flavors and fragrances
Documentation
see Safety & Documentation for available documents
food allergen
no known allergens
fragrance allergen
no known allergens
greener alternative category
Organoleptic
clean; creamy
SMILES string
CC(=O)OCC(COC(C)=O)OC(C)=O
InChI
1S/C9H14O6/c1-6(10)13-4-9(15-8(3)12)5-14-7(2)11/h9H,4-5H2,1-3H3
InChI key
URAYPUMNDPQOKB-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
280.4 °F - closed cup
Flash Point(C)
138 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
-
For your product W200712, Triacetin natural, wonder whether you have any certificates to prove its bio-carbon (also called as biogenic) content, for example C14 report. Do you confirm that this is 100% biobased?
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service