538728
Triethyl phosphate
ReagentPlus®, ≥99.8%
Synonym(s):
TEP, Ethyl phosphate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
(C2H5O)3PO
CAS Number:
Molecular Weight:
182.15
Beilstein:
1705772
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
vapor density
6.28 (vs air)
vapor pressure
1 mmHg ( 40 °C)
product line
ReagentPlus®
Assay
≥99.8%
autoignition temp.
845 °F
expl. lim.
10 %
refractive index
n20/D 1.403 (lit.)
bp
215 °C (lit.)
density
1.072 g/mL at 25 °C (lit.)
functional group
phosphate
SMILES string
CCOP(=O)(OCC)OCC
InChI
1S/C6H15O4P/c1-4-8-11(7,9-5-2)10-6-3/h4-6H2,1-3H3
InChI key
DQWPFSLDHJDLRL-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Triethyl phosphate is an alkyl phosphate. It participates in the thermal transformation of sodium acetate-2-C14 to n-butyl acetate-2-C14.
Application
- Sensor Technology for Chemical Detection: Triethyl phosphate is utilized in the development of trisaryl phosphoric triamide-based resins for a quartz crystal microbalance sensor, enhancing the detection capabilities for sulfur mustard simulants, which is critical in chemical safety and environmental monitoring (Heo et al., 2024).
- Flame-Retardant Bio-based Composites: Research highlights the use of triethyl phosphate in creating flame-retardant, energy-storage, and transparent bio-based composites, leveraging natural cellulose templates to enhance both safety and environmental sustainability (Li et al., 2024).
- Environmental Biodegradation Studies: Triethyl phosphate is instrumental in a novel method for rapidly monitoring organophosphate flame retardants (OPFRs) by liquid-liquid extraction and gas chromatography-mass spectrometry, a significant advancement in environmental science for assessing biodegradation (Losantos et al., 2024).
Legal Information
ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
266.0 °F - closed cup
Flash Point(C)
130 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Preparation of Ethyl Acetate-2-C14 and n-Butyl Acetate-2-C14 Using Alkyl Phosphates1
Ropp GA.
Journal of the American Chemical Society, 72.5 , 2299-2299 (1950)
Novel PVDF-HFP flat sheet membranes prepared by triethyl phosphate (TEP) solvent for direct contact membrane distillation
Fadhil S, et al.
Chemical Engineering and Processing, 102, 16-26 (2016)
Xiaoyu Du et al.
Materials science & engineering. C, Materials for biological applications, 103, 109731-109731 (2019-07-28)
The fabrication of bone tissue engineering scaffolds with high osteogenic ability and favorable mechanical properties is of huge interest. In this study, a silk fibroin (SF) solution of 30 wt% was extracted from cocoons and combined with mesoporous bioactive glass (MBG)
Xiaodi Ren et al.
Proceedings of the National Academy of Sciences of the United States of America, 117(46), 28603-28613 (2020-11-05)
Functional electrolyte is the key to stabilize the highly reductive lithium (Li) metal anode and the high-voltage cathode for long-life, high-energy-density rechargeable Li metal batteries (LMBs). However, fundamental mechanisms on the interactions between reactive electrodes and electrolytes are still not
A novel approach to recycling of polyurethanes: chemical degradation of flexible polyurethane foams by triethyl phosphate
Troev K, et al.
Polymer, 41.19, 7017-7022 (2000)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service