Skip to Content
Merck
All Photos(1)

Key Documents

W200700

Sigma-Aldrich

Triacetin

99%, FCC, FG

Synonym(s):

1,2,3-Triacetoxypropane, 1,2,3-Triacetylglycerol, Glyceryl triacetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3COOCH2)2CHOCOCH3
CAS Number:
Molecular Weight:
218.20
FEMA Number:
2007
Beilstein:
1792353
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
E Number:
E 1518
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1333/2008 & 178/2002
FCC
FDA 21 CFR 117
FDA 21 CFR 184.1901

vapor density

7.52 (vs air)

Assay

99%

autoignition temp.

809 °F

expl. lim.

1.05 %, 189 °F
7.73 %, 215 °F

refractive index

n25/D 1.429-1.431 (lit.)

bp

258 °C (lit.)

mp

3 °C (lit.)

density

1.16 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

clean

SMILES string

CC(=O)OCC(COC(C)=O)OC(C)=O

InChI

1S/C9H14O6/c1-6(10)13-4-9(15-8(3)12)5-14-7(2)11/h9H,4-5H2,1-3H3

InChI key

URAYPUMNDPQOKB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Triacetin is a triester of glycerin and acetic acid that occurs naturally in papaya. It is mainly used as a synthetic flavoring agent in ice-creams, nonalcoholic beverages and baked goods.

Application


  • Glycerol Triacetate-Based Flame Retardant High-Temperature Electrolyte for the Lithium-Ion Battery.: This paper highlights the use of Triacetin as a flame retardant and high-temperature stable electrolyte component in lithium-ion batteries, enhancing safety and efficiency (Wu X et al., 2024).

  • Understanding the Interaction of Thermal, Rheological, and Mechanical Parameters Critical for the Processability of Polyvinyl Alcohol-Based Systems during Hot Melt Extrusion.: Triacetin is investigated for its effects on the processability of polyvinyl alcohol in hot melt extrusion, focusing on its role as a plasticizer to modify thermal and mechanical properties (Hess F et al., 2024).

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

298.4 °F - closed cup

Flash Point(C)

148 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Burdock GA
Encyclopedia of Food and Color Additives, 1, 2822-2822 (1997)
Fenaroli G
Fenaroli's Handbook of Flavor Ingredients, 542-542 (1971)
Yannai S
Dictionary of Food Compounds with CD-ROM: Additives, Flavors, and Ingredients, 7-7 (2012)
Huayin Pu et al.
Journal of agricultural and food chemistry, 59(10), 5738-5745 (2011-04-26)
An oral colon-targeting controlled release system based on resistant starch acetate (RSA) as a film-coating material was developed. The RSA was successfully synthesized, and its digestion resistibility could be improved by increasing the degree of substitution (DS), which was favorable
Seung Jun Choi et al.
Journal of agricultural and food chemistry, 57(23), 11349-11353 (2009-11-07)
Citral is widely used in the beverage, food, and fragrance industries for its characteristic flavor profile. However, it chemically degrades over time in aqueous solutions due to an acid-catalyzed reaction, which leads to loss of desirable flavor notes and formation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service