T60607
Triethyl orthopropionate
97%
Synonym(s):
1,1,1-Triethoxypropane
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CH3CH2C(OC2H5)3
CAS Number:
Molecular Weight:
176.25
Beilstein:
906798
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
97%
refractive index
n20/D 1.402 (lit.)
bp
155-160 °C (lit.)
density
0.876 g/mL at 25 °C (lit.)
SMILES string
CCOC(CC)(OCC)OCC
InChI
1S/C9H20O3/c1-5-9(10-6-2,11-7-3)12-8-4/h5-8H2,1-4H3
InChI key
FGWYWKIOMUZSQF-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Triethyl orthopropionate is commonly used in the Claisen rearrangement to construct new C-C bonds. It can react with pyrrole and chloroacetic acid to form 1,1,1- [tri-(pyrrol-2-yl)]propane.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
140.0 °F - closed cup
Flash Point(C)
60 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Acyclic stereoselection. 44. Diastereoselectivity in the ortho ester Claisen rearrangement of chiral propargylic alcohols. Use of. beta.-allenic esters as chiral methylmalonaldehyde synthons.
Henderson M A and Heathcock C H
The Journal of Organic Chemistry, 53(20), 4736-4745 (1988)
Stereoselectivity in the ortho ester Claisen rearrangements of the E and Z isomers of. gamma.-(1, 3-dioxan-4-yl) allyl alcohols.
Tadano K, et al.
The Journal of Organic Chemistry, 55(7), 2108-2113 (1990)
Reactions between pyrrole and orthoesters: preparation of tri-(pyrrol-2-yl) alkanes.
Reese C B and Yan H
Tetrahedron Letters, 42(32), 5545-5547 (2001)
The Johnson-Claisen rearrangement of 3-hydroxy-2-methylenealkanenitriles: Stereoselective synthesis of functionalized trisubstituted alkenes.
Basavaiah D and Pandiaraju S
Tetrahedron Letters, 36(5), 757-758 (1995)
Claisen rearrangement of (Z)-3-deoxy-3-C-[(hydroxymethyl) methylene]-1, 2: 5, 6-di-O-isopropylidene-. alpha.-D-ribo-hexofuranose with triethyl orthopropionate.
Tadano K, et al.
The Journal of Organic Chemistry, 54(5), 1223-1227 (1989)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service