Skip to Content
Merck
All Photos(3)

Key Documents

113808

Sigma-Aldrich

1,3-Dichlorobenzene

98%

Synonym(s):

m-Dichlorobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H4Cl2
CAS Number:
Molecular Weight:
147.00
Beilstein:
956618
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

5 mmHg ( 38.8 °C)

Assay

98%

form

liquid

refractive index

n20/D 1.546 (lit.)

bp

172-173 °C (lit.)

mp

−25-−22 °C (lit.)

solubility

H2O: soluble 1.23 mg/L
alcohol: soluble
diethyl ether: soluble

density

1.288 g/mL at 25 °C (lit.)

functional group

chloro

SMILES string

Clc1cccc(Cl)c1

InChI

1S/C6H4Cl2/c7-5-2-1-3-6(8)4-5/h1-4H

InChI key

ZPQOPVIELGIULI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1,3-Dichlorobenzene was used to study the effect of solvent vapor pressure on the vertical nanowire of C60 molecules. 1,3-Dichlorobenzene, a chlorinated aromatic hydrocarbon, can be detected in environmental samples using an advanced Fourier transform infrared spectroscopy-attenuated total reflectance (FTIR-ATR) sensor.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

152.6 °F - closed cup

Flash Point(C)

67.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rui Lu et al.
Scientific reports, 3, 2525-2525 (2013-08-29)
Chlorinated aliphatic hydrocarbons and chlorinated aromatic hydrocarbons (CHCs) are toxic and carcinogenic contaminants commonly found in environmental samples, and efficient online detection of these contaminants is still challenging at the present stage. Here, we report an advanced Fourier transform infrared
Carina Dettenrieder et al.
Sensors (Basel, Switzerland), 19(17) (2019-08-24)
A portable sensor system for the simultaneous detection of multiple environmentally relevant volatile organic compounds (VOCs) in real seawater based on Fourier transform infrared fiber-optic evanescent wave spectroscopy (FT-IR-FEWS) was developed. A cylindrical silver halide (AgX) fiber with an ethylene/propylene
Jungah Kim et al.
ACS nano, 7(10), 9122-9128 (2013-09-11)
We report that C60 molecules are spontaneously crystallized into vertical nanowires by the solvent vapor annealing (SVA) process. C60 molecules have been known to be assembled into wire-like crystals by simply dropping and drying C60 solutions in m-xylene on a
N Yoshida et al.
Analytical chemistry, 73(18), 4417-4421 (2001-09-29)
A distributed-feedback dye laser has been developed for achieving the efficient multiphoton ionization of chlorobenzene and dichlorobenzene, that is, precursor molecules of dioxins. This tunable picosecond laser with a narrow spectral line width, that is, a nearly transform-limited pulse, provides
Claudia Amorim et al.
Journal of hazardous materials, 211-212, 208-217 (2011-08-30)
Catalytic hydrodechlorination (HDC) is an effective means of detoxifying chlorinated waste. Involvement of spillover hydrogen is examined in gas phase dechlorination of chlorobenzene (CB) and 1,3-dichlorobenzene (1,3-DCB) over Pd and Ni. The catalytic action of single component Pd and Ni

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service