Skip to Content
Merck
All Photos(3)

Key Documents

T400

Sigma-Aldrich

DL-Tartaric acid

ReagentPlus®, 99%

Synonym(s):

DL-2,3-Dihydroxybutanedioic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOOC(CHOH)2COOH
CAS Number:
Molecular Weight:
150.09
Beilstein:
1725148
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

product line

ReagentPlus®

Assay

99%

mp

210-212 °C (lit.)

SMILES string

O[C@@H]([C@H](O)C(O)=O)C(O)=O

InChI

1S/C4H6O6/c5-1(3(7)8)2(6)4(9)10/h1-2,5-6H,(H,7,8)(H,9,10)/t1-,2-/m0/s1

InChI key

FEWJPZIEWOKRBE-LWMBPPNESA-N

Looking for similar products? Visit Product Comparison Guide

Application

DL-Tartaric acid can be used:
  • In the Debus–Radziszewski reaction as a weak acid for the synthesis of imidazolium ionic liquid.
  • As an additive in electrochemical deposition technique for the synthesis of bismuth thin films to be used as X-ray absorbers.
  • As a complexing agent for the synthesis of nano-crystalline indium tin oxide (ITO) powder.
  • As a dopant for the synthesis of polyaniline nanofibers and nanotubes by oxidation polymerization.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

410.0 °F - closed cup

Flash Point(C)

210 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Green Imidazolium Ionics-From Truly Sustainable Reagents to Highly Functional Ionic Liquids.
Troger-M?ller S, et al.
Chemistry?A European Journal , 23(49), 11810-11817 (2017)
The unique role of DL-tartaric acid in determining the morphology of polyaniline nanostructures during an interfacial oxidation polymerization.
Sun Q and Deng Y
Materials Letters, 62(12-13), 1831-1834 (2008)
Transparent and conducting ITO thin films by spin coating of an aqueous precursor solution.
Sunde TOL, et al.
Journal of Materials Chemistry, 22(31), 15740-15749 (2012)
Microstructure and electrical transport in electrodeposited Bi films.
Moral-Vico J, et al.
Journal of Electroanalytical Chemistry, 832(49), 40-47 (2019)
Andrea Bencini et al.
Chemical communications (Cambridge, England), 48(84), 10428-10430 (2012-09-18)
A chiral ditopic polyammonium receptor featuring two [9]aneN(3) moieties separated by a (S)-BINOL linker is able to selectively bind and sense in water (S,S)-tartaric acid over its (R,R)/meso forms.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service