All Photos(3)
About This Item
Linear Formula:
(CH3)2CHCH(CH3)2
CAS Number:
Molecular Weight:
86.18
Beilstein:
1730737
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
vapor density
3 (vs air)
Quality Level
vapor pressure
4.54 psi ( 25.0 °C)(lit.)
Assay
98%
form
liquid
autoignition temp.
788 °F
expl. lim.
~7.7 %
refractive index
n20/D 1.375 (lit.)
bp
58 °C (lit.)
mp
−129 °C (lit.)
density
0.662 g/mL at 25 °C (lit.)
SMILES string
CC(C)C(C)C
InChI
1S/C6H14/c1-5(2)6(3)4/h5-6H,1-4H3
InChI key
ZFFMLCVRJBZUDZ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
2,3-Dimethylbutane can can undergo hydrogenolysis over supported catalysts of ruthenium, nickel, cobalt, and iron to form smaller hydrocarbons, mainly methane.
Application
- Synthesis, structure, and properties of the condensation product of hexaoxocyclohexane with 2, 3-bis (hydroxyamino)-2, 3-dimethylbutane: Discusses the chemical synthesis and structural analysis of a novel compound derived from hexaoxocyclohexane and 2,3-dimethylbutane (EV Tretyakov, AO Tkacheva, 2014).
- Catalysts by design - Surface organometallic chemistry on oxide and metal for dehydrogenation of 2, 3-dimethylbutane: This paper details the development of a selective catalyst system for the dehydrogenation of 2,3-dimethylbutane, emphasizing the stability and efficiency of the catalyst (P Rouge, A Garron, S Norsic, C Larabi, N Merle, 2019).
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
-20.2 °F - closed cup
Flash Point(C)
-29 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Gas hydrate equilibria of 2, 3-Dimethylbutane and benzene with methane and nitrogen.
?stergaard KK, et al.
Chemical Engineering Research and Design, 78(5), 731-737 (2000)
Kinetic study of the photochlorination of 2, 3-dimethylbutane and other alkanes in solution in the presence of benzene. First measurements of the absolute rate constants for hydrogen abstraction by the" free" chlorine atom and the chlorine atom-benzene. pi.-complex. Identification of these two species as the only hydrogen abstractors in these systems.
Bunce NJ, et al.
Journal of the American Chemical Society, 107(19), 5464-5472 (1985)
Hydrogenolysis of 2,3-dimethylbutane on supported ruthenium, nickel, cobalt, and iron.
Machiels CJ & Anderson RB
J. Catal., 58(2), 260-267 (1979)
Diffusion of benzene, toluene, naphthalene, and phenanthrene in supercritical dense 2, 3?dimethylbutane.
Sun CKJ & Chen SH
AIChE Journal, 31(11), 1904-1910 (1985)
Kinetics of C~ 6H~ 5 Radical Reactions with 2-Methylpropane, 2, 3-Dimethylbutane and 2, 3, 4-Trimethylpentane.
Park J, et al.
International Journal of Chemical Kinetics, 31(9), 645-653 (1999)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service