Skip to Content
Merck
All Photos(3)

Key Documents

L3609

Sigma-Aldrich

2,4-Lutidine

99%

Synonym(s):

2,4-Dimethylpyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H9N
CAS Number:
Molecular Weight:
107.15
Beilstein:
1506
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
liquid
Assay:
99%

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.499 (lit.)

bp

159 °C (lit.)

mp

−60 °C (lit.)

density

0.927 g/mL at 25 °C (lit.)

SMILES string

Cc1ccnc(C)c1

InChI

1S/C7H9N/c1-6-3-4-8-7(2)5-6/h3-5H,1-2H3

InChI key

JYYNAJVZFGKDEQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

116.6 °F - closed cup

Flash Point(C)

47 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Umesh Kumar et al.
Inorganic chemistry, 49(1), 62-72 (2009-12-05)
The reactions of Zn(OAc)(2).2H(2)O with various positional isomers of lutidine were explored with a view to understand the factors responsible for the nuclearity/aggregation and acetate coordination modes of the products. The reactions of Zn(OAc)(2).2H(2)O with 3,5-lutidine, 2,3-lutidine, 2,4-lutidine, and 3,4-lutidine
Roberto Fernández-Maestre et al.
The Analyst, 135(6), 1433-1442 (2010-04-07)
In ion mobility spectrometry (IMS), reduced mobility values (K(0)) are used as a qualitative measure of gas phase ions, and are reported in the literature as absolute values. Unfortunately, these values do not always match with those collected in the
J M Robert et al.
Arzneimittel-Forschung, 47(5), 635-642 (1997-05-01)
Derivatives of N-(4,6-pyridin-2-yl)arylcarboxamides resulting from the integration of the amidic function into 4H-1,2,4-triazole, triazol-3(2H)-one and 1H-tetrazole rings were evaluated as potential anti-inflammatory compounds. The level of activity decreased as compared to carboxamides, nevertheless their precursors and notably the corresponding amidrazones
S R Agapova et al.
Molekuliarnaia genetika, mikrobiologiia i virusologiia, (5-6)(5-6), 10-13 (1992-05-01)
Arthrobacter crysallopoietes strain KM-4 degrading 2,6-dimethylpyridine and strain KM-4a degrading both 2,6-dimethylpyridine and pyridine, Arthrobacter sp. KM-4b degrading 2,4-dimethylpyridine were isolated from soil. Arthrobacter crystallopoietes KM-4 and Arthrobacter sp. KM-4b contain 100 Md plasmids pBS320 and pBS323. Arthrobacter crystallopietes KM-4a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service