Skip to Content
Merck
All Photos(1)

Key Documents

W354007

Sigma-Aldrich

2,6-Dimethylpyridine

≥99%

Synonym(s):

2,6-Lutidine, 2,6-Dimethylpyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H9N
CAS Number:
Molecular Weight:
107.15
FEMA Number:
3540
Beilstein:
105690
EC Number:
Council of Europe no.:
11381
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
14.065
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

Halal
Kosher

Agency

meets purity specifications of JECFA

Assay

≥99%

refractive index

n20/D 1.497 (lit.)

bp

143-145 °C (lit.)

mp

−6 °C (lit.)

density

0.92 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

bread; nutty

SMILES string

Cc1cccc(C)n1

InChI

1S/C7H9N/c1-6-4-3-5-7(2)8-6/h3-5H,1-2H3

InChI key

OISVCGZHLKNMSJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

2,6-Dimethylpyridine is a volatile flavor compound that is reported to occur in soy sauce and boiled buckwheat flour. It is also one of the key volatiles bases found in cigar and cigarette smoke.

Application


  • Study on the correlation between the dominant microflora and the main flavor substances in the fermentation process of cigar tobacco leaves.: Research examines the role of 2,6-Dimethylpyridine in tobacco fermentation, highlighting its impact on flavor development and microbial activity, which is crucial for optimizing product quality in the tobacco industry (Wu et al., 2023).

  • The Impact of ortho-substituents on Bonding in Silver(I) and Halogen(I) Complexes of 2-Mono- and 2,6-Disubstituted Pyridines: An In-Depth Experimental and Theoretical Study.: This investigation details how modifications like 2,6-dimethylpyridine influence bonding and reactivity in metal complexes, providing insights that could influence the design of new catalytic systems or electronic materials (Kumar et al., 2024).

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

89.6 °F

Flash Point(C)

32 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shoyu (soy sauce) volatile flavor components: basic fraction.
Nunomura N, et al.
Agricultural and Biological Chemistry, 42(11), 2123-2128 (1978)
Chemical studies on tobacco smoke. LXI. Volatile pyridines: Quantitative analysis in mainstream and sidestream smoke of cigarettes and cigars.
Brunnemann KD, et al.
Analytical Letters, 11(7), 545-560 (1978)
The volatile bases of cigar smoke.
Osman S & Barson J.
Phytochemistry, 3(5), 587-590 (1964)
Volatile flavor compounds of boiled buckwheat flour
Yajima I, et al.
Agricultural and Biological Chemistry, 47(4), 729-738 (1983)
Bojana Ginovska et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 21(44), 15713-15719 (2015-10-24)
We report that 2,6-lutidine⋅trichloroborane (Lut⋅BCl3 ) reacts with H2 in toluene, bromobenzene, dichloromethane, and Lut solvents producing the neutral hydride, Lut⋅BHCl2 . The mechanism was modeled with density functional theory, and energies of stationary states were calculated at the G3(MP2)B3

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service