Skip to Content
Merck
All Photos(3)

Documents

161683

Sigma-Aldrich

4-Cyclopentene-1,3-dione

95%

Synonym(s):

1-Cyclopentene-3,5-dione, 2-Cyclopenten-1,4-dione, 2-Cyclopentene-1,4-dione, Cyclopent-4-en-1,3-dione, Cyclopentene-3,5-dione, Cyclopentenedione

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C5H4(=O)2
CAS Number:
Molecular Weight:
96.08
Beilstein:
2038507
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

bp

60 °C/1 mmHg (lit.)

mp

34-36 °C (lit.)

storage temp.

2-8°C

SMILES string

O=C1CC(=O)C=C1

InChI

1S/C5H4O2/c6-4-1-2-5(7)3-4/h1-2H,3H2

InChI key

MCFZBCCYOPSZLG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

4-Cyclopentene-1,3-dione was used in the synthesis of 2-methoxymethylene-4-cyclopentene-1,3-dione and chrysotrione B (2-acylcyclopentene-1,3-dione derivative). It was used to investigate the influence of sulfiydryl group-specific reagents on activity of bovine heart phospholipid exchange protein.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

183.2 °F - closed cup

Flash Point(C)

84 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P E DiCorleto et al.
The Journal of biological chemistry, 254(16), 7795-7802 (1979-08-25)
Two phospholipid exchange proteins from bovine heart have been purified approximately 2000-fold and judged greater than 90% pure. The proteins are similar in molecular weight (both 33,400 by polyacrylamide gel electrophoresis and 23,500 by gel filtration), in amino acid composition
Yoshihiro Uto et al.
Bioorganic & medicinal chemistry, 16(11), 6042-6053 (2008-05-14)
We designed chiral 2-nitroimidazole derivatives containing a 2-aminomethylene-4-cyclopentene-1,3-dione moiety as antiangiogenic hypoxic cell radiosensitizers. Based on results of molecular orbital calculations, the 2-aminomethylene-4-cyclopentene-1,3-dione moiety is expected to show high electrophilicity comparable to that of the 2-methylene-4-cyclopentene-1,3-dione moiety included in TX-1123
T Bui et al.
Biochimica et biophysica acta, 1397(1), 31-42 (1998-04-18)
The molecular pathways by which the cyclopentenone prostaglandins (PGA and PGJ series) inhibit cell growth and tumorigenicity are poorly understood. These cellular responses may be caused by specific regulation of growth-related and stress-induced genes. A variety of prostaglandins were tested
Mona Mokhtari et al.
Journal of agricultural and food chemistry, 66(22), 5531-5539 (2018-03-17)
Pathogenic fungi continue to develop resistance against current antifungal drugs. To explore the potential of agricultural waste products as a source of novel antifungal compounds, we obtained an unbiased GC-MS profile of 151 compounds from 16 commercial and experimental cultivars
Jiang Qian et al.
Chemical communications (Cambridge, England), 47(32), 9203-9205 (2011-07-09)
Facile, two-step synthesis and kinetic characterization of new chemical probes for selective labeling of sulfenic acid (-SOH) in proteins are presented. The synthesis route relies on the simple and highly efficient Michael addition of thiol containing tags or linkers to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service