Skip to Content
Merck
All Photos(1)

Key Documents

515981

Sigma-Aldrich

1,3-Cycloheptanedione

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C7H10(=O)2
CAS Number:
Molecular Weight:
126.15
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

refractive index

n20/D 1.483 (lit.)

bp

254 °C (lit.)

density

1.1 g/mL at 25 °C (lit.)

functional group

ketone

SMILES string

O=C1CCCCC(=O)C1

InChI

1S/C7H10O2/c8-6-3-1-2-4-7(9)5-6/h1-5H2

InChI key

DBOVMTXPZWVYAQ-UHFFFAOYSA-N

Related Categories

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Allen Y Hong et al.
Tetrahedron, 67(52), 10234-10248 (2012-02-22)
General catalytic asymmetric routes toward cyclopentanoid and cycloheptanoid core structures embedded in numerous natural products have been developed. The central stereoselective transformation in our divergent strategies is the enantioselective decarboxylative alkylation of seven-membered β-ketoesters to form α-quaternary vinylogous esters. Recognition
Michael additions of 1, 3-cycloalkanediones to dimethyl acety-lenedicarboxylate.
Hrnciar P, et al.
Chemical Papers, 43(1), 87-95 (1989)
Methylation of 1, 3-cyclopentanedione, 1, 3-cyclohexanedione, and 1, 3-cycloheptanedione with iodomethane in aprotic solvents in the absence and in the presence of 18-crown-6.
Sraga J and Hrnciar P.
Chemical Papers, 35(1), 119-126 (1981)
Evan A Sims et al.
Tetrahedron letters, 52(16), 1871-1873 (2011-06-29)
We report the large-scale synthesis of 1,3-cyclooctanedione in five steps with 29% yield. This molecule is a synthetic precurser to difluorinated cyclooctyne, which participates in a bioorthogonal copper-free click reaction with azides. The final step demonstrates the first successful application
Michael addition of 1, 3-cyclopentanedione, 1, 3-cyclohexanedione and 1, 3-cycloheptanedione to 1-(X-phenyl)-2-nitroethylenes.
Hrnciar P and Culak I.
Collection of Czechoslovak Chemical Communications, 49(6), 1421-1431 (1984)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service