Skip to Content
Merck
All Photos(2)

Key Documents

GF88601531

Gadolinium

rod, 100mm, diameter 3.0mm, cast, 99%

Synonym(s):

Gadolinium, GD007905

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Gd
CAS Number:
Molecular Weight:
157.25
MDL number:
UNSPSC Code:
12141605
PubChem Substance ID:
NACRES:
NA.23

Assay

≥99%

form

rod

manufacturer/tradename

Goodfellow 886-015-31

resistivity

126 μΩ-cm, 20°C

bp

3273 °C (lit.)

mp

1313 °C (lit.)

density

7.886 g/mL at 25 °C (lit.)

SMILES string

[Gd]

InChI

1S/Gd

InChI key

UIWYJDYFSGRHKR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Water-react 3

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Brigit den Adel et al.
Atherosclerosis, 225(2), 274-280 (2012-08-14)
MRI using targeted contrast agents (CA) has emerged as a promising technique to study atherothrombotic disease in vivo. Particularly, the use of targeted Gd and lipid-based nanoparticles has enabled detailed in vivo imaging of various molecular markers of atherosclerotic plaque
Kanae Kawai Miyake et al.
AJR. American journal of roentgenology, 203(2), 272-279 (2014-07-24)
The purpose of this study was to assess the diagnostic value of (18)F-FDG PET/CT and MRI in predicting the clinicopathologic subtypes of breast cancer. The cases of 89 patients with mass-type invasive breast cancer who underwent FDG PET/CT and MRI
Giovanni Antonini et al.
Pain, 155(8), 1464-1471 (2014-05-03)
Although classical trigeminal neuralgia (CTN) is frequently caused by neurovascular contact (NVC) at the trigeminal root entry zone (REZ), both anatomical and MRI studies have shown that NVC of the trigeminal nerve frequently occurs in individuals without CTN. To assess
Tevfik F Ismail et al.
Heart (British Cardiac Society), 100(23), 1851-1858 (2014-06-27)
Myocardial fibrosis identified by late gadolinium enhancement (LGE) cardiovascular magnetic resonance (CMR) in patients with hypertrophic cardiomyopathy (HCM) is associated with adverse cardiovascular events, but its value as an independent risk factor for sudden cardiac death (SCD) is unknown. We
John J Green et al.
JACC. Cardiovascular imaging, 5(4), 370-377 (2012-04-14)
The objective of this study was to perform a systematic review and meta-analysis of the predictive value of late gadolinium enhancement (LGE) cardiac magnetic resonance (CMR) for future cardiovascular events and death in hypertrophic cardiomyopathy (HCM). The utility of LGE

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service