Recommended Products
Assay
99.9% trace rare earth metals basis
form
chips
reaction suitability
reagent type: catalyst
core: gadolinium
resistivity
126 μΩ-cm, 20°C
bp
3273 °C (lit.)
mp
1313 °C (lit.)
density
7.886 g/mL at 25 °C (lit.)
SMILES string
[Gd]
InChI
1S/Gd
InChI key
UIWYJDYFSGRHKR-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Water-react 3
Storage Class Code
4.3 - Hazardous materials which set free flammable gases upon contact with water
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service