Skip to Content
Merck
All Photos(2)

Key Documents

263060

Sigma-Aldrich

Gadolinium

−40 mesh, 99% trace rare earth metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Gd
CAS Number:
Molecular Weight:
157.25
EC Number:
MDL number:
UNSPSC Code:
12141605
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99% trace rare earth metals basis

form

powder

reaction suitability

reagent type: catalyst
core: gadolinium

resistivity

126 μΩ-cm, 20°C

particle size

−40 mesh

bp

3273 °C (lit.)

mp

1313 °C (lit.)

density

7.886 g/mL at 25 °C (lit.)

SMILES string

[Gd]

InChI

1S/Gd

InChI key

UIWYJDYFSGRHKR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Water-react 3

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Stephen Patrick et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(1), 415-420 (2013-12-19)
The ability to track cells and their patterns of gene expression in living organisms can increase our understanding of tissue development and disease. Gene reporters for bioluminescence, fluorescence, radionuclide, and magnetic resonance imaging (MRI) have been described but these suffer
Tomas G Neilan et al.
Journal of the American College of Cardiology, 62(23), 2205-2214 (2013-09-03)
This study sought to identify the frequency, pattern, and prognostic significance of left ventricular (LV) late gadolinium enhancement (LGE) in patients with atrial fibrillation (AF). There are limited data on the presence, pattern, and prognostic significance of LV myocardial fibrosis
Yusuke Nishimura et al.
Journal of neurosurgery. Spine, 19(6), 716-720 (2013-10-08)
A spinal intradural extramedullary venous angioma is extremely rare and has not been previously reported. In this paper, the authors report on this entity with morphological and immunohistochemical evidence, and discuss the surgical strategy for its treatment. A 54-year-old woman
Vanessa Wiggermann et al.
Neurology, 81(3), 211-218 (2013-06-14)
We investigated the evolution of new multiple sclerosis (MS) lesions over time using frequency shifts of the magnetic resonance (MR) signal. Twenty patients with relapsing-remitting MS were serially scanned for 6 months at 1-month intervals. Maps of MR frequency shifts
Stéphane Kremer et al.
Neurology, 81(3), 206-210 (2013-06-14)
The first goal of this study is to compare gadofosveset trisodium--a gadolinium agent that reversibly binds to albumin--to an extracellular contrast agent (Gd-DOTA) for the detection of multiple sclerosis lesions. The second goal is to determine the best postinjection time

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service