Skip to Content
Merck
All Photos(2)

Documents

GF66395212

Selenium

foil, 25x25mm, thickness 3mm, 99.95%

Synonym(s):

Selenium, SE000500

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Se
CAS Number:
Molecular Weight:
78.96
MDL number:
UNSPSC Code:
12141910
PubChem Substance ID:
NACRES:
NA.23

description

gray

Assay

99.95%

form

foil

manufacturer/tradename

Goodfellow 663-952-12

resistivity

1.2 μΩ-cm, 0°C

size × thickness

25x25 mm × 3.0 mm

bp

684.9 °C (lit.)

mp

217 °C (lit.)

density

4.81 g/mL at 25 °C (lit.)

SMILES string

[Se]

InChI

1S/Se

InChI key

BUGBHKTXTAQXES-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 4 - STOT RE 2

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kai-Oliver Feldmann et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 21(27), 9697-9712 (2015-05-12)
Although a fairly large number of binary group 15/16 element cations have been reported, no example involving phosphorus in combination with a group 16 element has been synthesized and characterized to date. In this contribution is reported the synthesis and structural characterization
Claire M Weekley et al.
Nutrients, 5(5), 1734-1756 (2013-05-24)
Determining the speciation of selenium in vivo is crucial to understanding the biological activity of this essential element, which is a popular dietary supplement due to its anti-cancer properties. Hyphenated techniques that combine separation and detection methods are traditionally and
Matthew Loxham et al.
Toxicological sciences : an official journal of the Society of Toxicology, 145(1), 98-107 (2015-02-13)
We have previously shown that underground railway particulate matter (PM) is rich in iron and other transition metals across coarse (PM10-2.5), fine (PM2.5), and quasi-ultrafine (PM0.18) fractions and is able to generate reactive oxygen species (ROS). However, there is little
Mark Denham et al.
Stem cells (Dayton, Ohio), 33(6), 1759-1770 (2015-03-11)
The caudal neural plate is a distinct region of the embryo that gives rise to major progenitor lineages of the developing central and peripheral nervous system, including neural crest and floor plate cells. We show that dual inhibition of the
Joseph L Jacobson et al.
Environmental health perspectives, 123(8), 827-833 (2015-03-11)
Although prenatal methylmercury exposure has been linked to poorer intellectual function in several studies, data from two major prospective, longitudinal studies yielded contradictory results. Associations with cognitive deficits were reported in a Faroe Islands cohort, but few were found in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service