Skip to Content
Merck
All Photos(2)

Key Documents

GF34805268

Cobalt

foil, 25x25mm, thickness 1.0mm, as rolled, 99.99+%

Synonym(s):

Cobalt, CO000291

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Co
CAS Number:
Molecular Weight:
58.93
MDL number:
UNSPSC Code:
12141710
PubChem Substance ID:
NACRES:
NA.23

Assay

99.99%

form

foil

manufacturer/tradename

Goodfellow 348-052-68

resistivity

6.24 μΩ-cm, 20°C

size × thickness

25x25 mm × 1.0 mm

bp

2900 °C (lit.)

density

8.9 g/mL at 25 °C (lit.)

SMILES string

[Co]

InChI

1S/Co

InChI key

GUTLYIVDDKVIGB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 1B - Eye Irrit. 2 - Muta. 2 - Repr. 1A - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dalia Nayak et al.
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine, 65(8), 891-896 (2007-05-29)
Heavy ion activation of natural cobalt foil with 84MeV (12)C results in the formation of no-carrier-added (nca) (66,67)As radionuclides, along with their corresponding decay products, (66,67)Ge and (66,67)Ga, in the matrix. Because arsenic and germanium radionuclides are short-lived, after a
M D Wirt et al.
Biochemistry, 34(15), 5269-5273 (1995-04-18)
We have completed the first direct structural characterization of an enzyme-bound four-coordinate Co(I) intermediate, in this case for the corrinoid/iron-sulfur protein (C/Fe-SP) from Clostridium thermoaceticum. Extended X-ray absorption fine structure and X-ray edge spectroscopy of the active Co(I) state of
Kian Keat Lee et al.
Biosensors & bioelectronics, 39(1), 255-260 (2012-08-18)
Cobalt oxyhydroxide, CoOOH, nanosheets were prepared via a surface alkaline treatment of cobalt foil at room temperature without using templates and catalysts. The morphology, chemical composition and structures of the nanosheets were characterized by XRD, FTIR and Raman spectroscopy, FESEM
D Nayak et al.
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine, 54(2), 189-193 (2001-02-24)
Heavy ion activation of natural cobalt foil with 52 MeV 11B4+ and 73 MeV 12C6+ results in the formation of carrier-free (66,67)Ge and (66,67)As radionuclides, respectively, along with their corresponding decay products, (66,67)Ga in the matrix. The carrier free gallium
Kenneth M Unice et al.
Chemico-biological interactions, 216, 53-74 (2014-04-15)
An updated biokinetic model for human exposures to cobalt (Co) was developed based on a comprehensive set of human pharmacokinetics data collected from five male and five female volunteers who ingested ∼1 mg Co/day of a Co supplement for 3

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service