Skip to Content
Merck
All Photos(6)

Key Documents

F7378

Sigma-Aldrich

Fluorescein diacetate

Powder

Synonym(s):

3,6-Diacetoxyfluoran, Di-O-acetylfluorescein

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H16O7
CAS Number:
Molecular Weight:
416.38
Beilstein:
62575
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Product Name

Fluorescein diacetate, used as cell viability stain

form

powder

technique(s)

titration: suitable

color

white to faint yellow

mp

200-203 °C (lit.)

solubility

acetone: 25 mg/mL

ε (extinction coefficient)

≥86000 at 488-494 nm in 0.1 M NaOH

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

CC(=O)Oc1ccc2c(Oc3cc(OC(C)=O)ccc3C24OC(=O)c5ccccc45)c1

InChI

1S/C24H16O7/c1-13(25)28-15-7-9-19-21(11-15)30-22-12-16(29-14(2)26)8-10-20(22)24(19)18-6-4-3-5-17(18)23(27)31-24/h3-12H,1-2H3

InChI key

CHADEQDQBURGHL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Fluorescein diacetate (FDA) is a non-fluorescent molecule, which is hydrolysed to fluorescent fluorescein in live cells. Dead cells cannot accumulate or hydrolyse FDA.

Application

Fluorescein diacetate has been used for the differentiation of live cells from dead cells.

Substrates

Lipase substrate

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The influence of scaffold microstructure on chondrogenic differentiation of mesenchymal stem cells.
Zhang J
Biomedical Materials (Bristol, England), 9, 035011-035011 (2014)
Effect of shock waves on suspended and immobilized L1210 cells.
Brummer F
Ultrasound in Medicine & Biology, 15, 229-229 (1989)
Cell Growth, Differentiation and Senescence: A Practical Approach (1999)
Koichi Baba et al.
Journal of controlled release : official journal of the Controlled Release Society, 153(3), 278-287 (2011-05-10)
This report describes a method for enhancing the ocular penetration of eye drops using nanoparticles of hydrolyzable dye, which is similar to a prodrug approach. The entry of eye drops into the ocular globe is restricted predominantly by corneal barrier
Derek Toms et al.
Frontiers in bioengineering and biotechnology, 8, 534-534 (2020-06-26)
We have developed an accessible software tool (receptoR) to predict potentially active signaling pathways in one or more cell type(s) of interest from publicly available transcriptome data. As proof-of-concept, we applied it to mouse photoreceptors, yielding the previously untested hypothesis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service