Skip to Content
Merck
All Photos(1)

Key Documents

E4402

Sigma-Aldrich

2-Ethoxybenzamide

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5OC6H4CONH2
CAS Number:
Molecular Weight:
165.19
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

mp

132-134 °C (lit.)

SMILES string

CCOc1ccccc1C(N)=O

InChI

1S/C9H11NO2/c1-2-12-8-6-4-3-5-7(8)9(10)11/h3-6H,2H2,1H3,(H2,10,11)

InChI key

SBNKFTQSBPKMBZ-UHFFFAOYSA-N

Related Categories

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yoshihiro Hayashi et al.
Pharmaceutics, 12(7) (2020-07-02)
We previously reported a novel method for the precise prediction of tablet properties (e.g., tensile strength (TS)) using a small number of experimental data. The key technique of this method is to compensate for the lack of experimental data by
N Hirasawa et al.
Chemical & pharmaceutical bulletin, 47(3), 417-420 (1999-04-23)
Solid dispersions of carbamazepine or ethenzamide were prepared by melting and rapid cooling with liquid nitrogen using lactose as a carrier. The physical characteristics of these solid dispersions were investigated by powder X-ray diffraction, differential scanning calorimetry, and dissolution rate
Y Miyamoto et al.
Chemical & pharmaceutical bulletin, 46(9), 1432-1437 (1998-10-17)
A computer optimization technique based on surface response methodology was applied to optimize the wet granulation process for designing tablets. Physical properties (mean granule size, granule size distribution, compressibility, granule strength) of a model granule formulation containing ethenzamide were accurately
Tadashi Fukunaka et al.
Journal of pharmaceutical sciences, 94(5), 1004-1012 (2005-03-29)
Milling is a common procedure to improve bioavailability of many active pharmaceutical ingredients (APIs), which typically have low solubility in water. But such micronization can yield an increase in the cohesiveness of particles. Although particle cohesiveness is desirable for tablet
Kunikazu Moribe et al.
Chemical & pharmaceutical bulletin, 52(5), 524-529 (2004-05-11)
We prepared and characterized a grinding-induced equimolar complex of thiourea with ethenzamide. When thiourea and ethenzamide were co-ground at a molar ratio of 3 : 1, new powder X-ray diffraction (PXRD) peaks were observed in addition to PXRD peaks of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service