Skip to Content
Merck
All Photos(1)

Key Documents

W242209

Sigma-Aldrich

Ethyl benzoate

≥99%, FCC, FG

Synonym(s):

Benzoic acid ethyl ester, Benzoic ether, Ethyl benzene carboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5COOC2H5
CAS Number:
Molecular Weight:
150.17
FEMA Number:
2422
Beilstein:
1908172
EC Number:
Council of Europe no.:
261
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.726
NACRES:
NA.21
grade:
FG
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117
FDA 21 CFR 172.515

vapor density

5.17 (vs air)

vapor pressure

1 mmHg ( 44 °C)

Assay

≥99%

autoignition temp.

914 °F

refractive index

n20/D 1.504 (lit.)

bp

212 °C (lit.)

mp

−34 °C (lit.)

density

1.045 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

anise; berry; cherry; balsamic; fruity; wintergreen

SMILES string

CCOC(=O)c1ccccc1

InChI

1S/C9H10O2/c1-2-11-9(10)8-6-4-3-5-7-8/h3-7H,2H2,1H3

InChI key

MTZQAGJQAFMTAQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Biochem/physiol Actions

Odor at 1.0%
Taste at 10 ppm

Other Notes

Natural occurrence: Feijoa, guava, plum, raspberry, rum, strawberry, Virginia tobacco.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

190.4 °F - closed cup

Flash Point(C)

88 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Organoleptic Characteristics of Flavor Materials
Mosciano, G.
Perfumer & Flavorist, 28, 102-102 (2003)
Daniel I Perez et al.
Bioorganic & medicinal chemistry, 17(19), 6914-6925 (2009-09-15)
Thienylhalomethylketones, whose chemical, biological, and pharmaceutical data are here reported, are the first irreversible inhibitors of GSK-3beta described to date. Their inhibitory activity is likely related to the cysteine residue present in the ATP-binding site, which is proposed as a
Alexandra Vinagre et al.
The journal of adhesive dentistry, 17(2), 107-116 (2015-04-29)
This study compared the microtensile bond strengths (μTBS) of two etch-and-rinse (ER) (OptiBond FL [OBFL]; Prime & Bond NT [PBNT]) and three self-etching (SE) (Clearfil SE Bond [CSEB]; Xeno III [XIII]; Xeno V+ [XV+]) adhesives systems to bur-prepared human enamel
Suvi-Katriina Ruokonen et al.
Journal of chromatography. A, 1405, 178-187 (2015-06-15)
The effect of three phosphonium and imidazolium ionic liquids (ILs) on the interaction between liposomes and common pharmaceuticals found in wastewaters was studied. The liposomes comprised zwitterionic phosphatidyl choline and negatively charged phosphatidyl glycerol. A set of common cationic, anionic
Katarzyna Niedojadło et al.
Plant cell reports, 34(12), 2201-2215 (2015-09-12)
Calreticulin expression is upregulated during sexual reproduction of Hyacinthus orientalis, and the protein is localized both in the cytoplasm and a highly specialized cell wall within the female gametophyte. Several evidences indicate calreticulin (CRT) as an important calcium (Ca(2+))-binding protein

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service