Skip to Content
Merck
All Photos(2)

Documents

D221309

Sigma-Aldrich

1,12-Dodecanediol

99%

Synonym(s):

Dodecamethylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO(CH2)12OH
CAS Number:
Molecular Weight:
202.33
Beilstein:
1742760
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

crystals

bp

189 °C/12 mmHg (lit.)

mp

79-81 °C (lit.)

SMILES string

OCCCCCCCCCCCCO

InChI

1S/C12H26O2/c13-11-9-7-5-3-1-2-4-6-8-10-12-14/h13-14H,1-12H2

InChI key

GHLKSLMMWAKNBM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

348.8 °F - closed cup

Flash Point(C)

176.00 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kangming Xu et al.
Polymers, 11(5) (2019-05-18)
Although hindered phenol/polymer-based hybrid damping materials, with excellent damping performance, attract more and more attention, the poor stability of hindered phenol limits the application of such promising materials. To solve this problem, a linear hindered phenol with amorphous state and
Aruuhan Bayaguud et al.
Dalton transactions (Cambridge, England : 2003), 46(14), 4602-4608 (2017-03-23)
A reaction strategy for the post-functionalization of hexavanadate derivatives is presented herein. In this study, five polyoxovanadate-based organic-inorganic hybrids TBA
Dina Maniar et al.
ChemSusChem, 12(5), 990-999 (2019-01-15)
Enzymatic polymerization provides an excellent opportunity for the conversion of renewable resources into polymeric materials in an effective and sustainable manner. A series of furan-based copolyesters was synthesized with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service