Skip to Content
Merck
All Photos(1)

Key Documents

674400

Sigma-Aldrich

1,16-Hexadecanedithiol

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H34S2
CAS Number:
Molecular Weight:
290.57
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

99%

form

solid

mp

48-52 °C

storage temp.

2-8°C

SMILES string

SCCCCCCCCCCCCCCCCS

InChI

1S/C16H34S2/c17-15-13-11-9-7-5-3-1-2-4-6-8-10-12-14-16-18/h17-18H,1-16H2

InChI key

JSRUFBZERGYUAT-UHFFFAOYSA-N

General description

1,16-Hexadecanedithiol (HDDT) is an alkanethiol that facilitates the formation of self-assembled monolayers (SAMs). It is majorly used to immobilize the surface atoms by attaching the thiol groups.

Application

HDDT forms a SAM on gold nanoparticle for potential usage in biosensors and other biological applications.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Journal of Physical Chemistry B, 107(30), 7406-7413 (2003)
Selective Nucleation and Area-Selective OMCVD of Gold on Patterned Self-Assembled Organic Monolayers Studied by AFM and XPS: A Comparison of OMCVD and PVD
Winter C, et al.
Chem. Vap. Deposition, 6(4), 199-205 (2000)
Templated growth of calcite, vaterite and aragonite crystals onself-assembled monolayers of substituted alkylthiols on gold
Kuther J, et al.
Journal of Materials Chemistry, 8(3), 641-650 (1998)
Aging of 1, omega-alkyldithiol interlinked au nanoparticle networks
Joseph Y, et al.
Chemistry of Materials, 21(8), 1670-1676 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service