Skip to Content
Merck
All Photos(3)

Key Documents

O3303

Sigma-Aldrich

1,8-Octanediol

98%

Synonym(s):

Octamethylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO(CH2)8OH
CAS Number:
Molecular Weight:
146.23
Beilstein:
1633499
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

bp

172 °C/20 mmHg (lit.)

mp

57-61 °C (lit.)

SMILES string

OCCCCCCCCO

InChI

1S/C8H18O2/c9-7-5-3-1-2-4-6-8-10/h9-10H,1-8H2

InChI key

OEIJHBUUFURJLI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1,8-Octanediol can undergo:
  • Polycondensation with citric acid to form biodegradable poly(1,8-octanediol citrate)(POC) crosslinked bioelastomer which can be blended with various additives.
  • Fischer esterification with dicarboxylic acids to form diol-based macromers.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

248.0 °F - closed cup

Flash Point(C)

120 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuanzheng Zhang et al.
PloS one, 12(8), e0182971-e0182971 (2017-08-12)
To explore the repairing effect of combination of adipose stem cells (ASCs) and composite scaffolds on CWR, the electrospun Poly 1, 8-octanediol-co-citric acid (POC)-poly-L-lactide acid (PLA) composite scaffolds were prepared, followed by in vitro and in vivo biocompatibility evaluation of
Yun He et al.
Acta biomaterialia, 93, 180-191 (2019-03-31)
The design and development of bioactive materials that are inherently conducive for osteointegration and bone regeneration with tunable mechanical properties and degradation remains a challenge. Herein, we report the development of a new class of citrate-based materials with glycerophosphate salts
Development of poly (1, 8-octanediol citrate)/chitosan blend films for tissue engineering applications.
Zeimaran E, et al.
Carbohydrate Polymers, 175, 618-627 (2017)
Kai-Hee Huong et al.
International journal of biological macromolecules, 116, 217-223 (2018-05-04)
Long carbon chain alkanediols are used in the production of poly(3-hydroxybutyrate-co-4-hydroxybutyrate) [P(3HB-co-4HB)], however these substrates possess high toxicity towards bacterial cells. This study demonstrated the effective utilisation of a long carbon chain alkanediol, namely 1,8-octanediol, to enhance the yield and
Synthesis and Characterization of Diol-Based Unsaturated Polyesters: Poly (diol fumarate) and Poly (diol fumarate-co-succinate).
Tatara AM, et al.
Biomacromolecules, 18(6), 1724-1735 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service