Skip to Content
Merck
All Photos(1)

Documents

D16401

Sigma-Aldrich

1,12-Diaminododecane

98%

Synonym(s):

1,12-Dodecanediamine, Dodecamethylenediamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2(CH2)12NH2
CAS Number:
Molecular Weight:
200.36
Beilstein:
1742765
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

flakes

mp

67-69 °C (lit.)

SMILES string

NCCCCCCCCCCCCN

InChI

1S/C12H28N2/c13-11-9-7-5-3-1-2-4-6-8-10-12-14/h1-14H2

InChI key

QFTYSVGGYOXFRQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Feedstock for polymer synthesis. Source of twelve carbon chain for medicinal drugs.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

311.0 °F - closed cup

Flash Point(C)

155 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kangkyun Baek et al.
Chemical communications (Cambridge, England), 46(23), 4091-4093 (2010-05-19)
The X-ray crystal structure of 1,12-dodecane diammonium (C(12)DA(2+)) encapsulated in cucurbit[8]uril reveals an unconventional U-shaped conformation of C(12)DA(2+), which is attributed to the favorable host-guest interactions to overcome the charge-charge repulsion of the two ammonium groups in close proximity and
Michelle Henderson Pozzi et al.
Biochemistry, 48(7), 1508-1516 (2009-02-10)
Mammalian polyamine oxidases (PAOs) catalyze the oxidation of N1-acetylspermine and N1-acetylspermidine to produce N-acetyl-3-aminopropanaldehyde and spermidine or putrescine. Structurally, PAO is a member of the monoamine oxidase family of flavoproteins. The effects of pH on the kinetic parameters of mouse
Monatshefte fur Chemie / Chemical Monthly, 124, 23-23 (1993)
E Dziembor-Gryszkiewicz et al.
Biochemistry international, 6(5), 627-633 (1983-05-01)
The acid phosphatase [EC 3.1.3.2] from human prostate gland is very unstable glycoprotein. To stabilize the enzyme cross-linking reaction with diamines was adopted. The carboxyl groups of the enzyme were activated with 1-ethyl-(3-dimethylaminopropyl)-carbodiimide and then treated with diamines of H2N-(CH2)n-NH2
Andrew R Hirst et al.
Langmuir : the ACS journal of surfaces and colloids, 20(25), 10851-10857 (2004-12-01)
The self-assembly of diaminododecane with dendritic l-lysine-based peptides to form gel-phase materials was investigated in a range of different solvents. The degree of structuring was modulated by the solvent employed, an effect which induced subtle changes in the mesoscale aggregate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service