Skip to Content
Merck
All Photos(3)

Documents

385506

Sigma-Aldrich

2,2′-(Ethylenedioxy)bis(ethylamine)

98%

Synonym(s):

1,2-Bis(2-aminoethoxy)ethane, 1,8-Diamino-3,6-dioxaoctane, 2,2′-(Ethylenedioxy)diethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CH2CH2OCH2CH2OCH2CH2NH2
CAS Number:
Molecular Weight:
148.20
Beilstein:
1739187
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

form

liquid

refractive index

n20/D 1.461 (lit.)

bp

105-109 °C/6 mmHg (lit.)

density

1.015 g/mL at 25 °C (lit.)

SMILES string

NCCOCCOCCN

InChI

1S/C6H16N2O2/c7-1-3-9-5-6-10-4-2-8/h1-8H2

InChI key

IWBOPFCKHIJFMS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2,2′-(ethylenedioxy)-bis-(ethylamine) (EDEA) was used to linker to form conjugates of:manganese-doped zinc sulfide nanoparticles with folic acid and dimeric neomycin-neomycin. Glycidyl groups of the l-GMA particles were reacted with EDEA to produce large amine-functionalized particles (l-NH2). Polycondensation of guanidine hydrochloride and 2,2′-(ethylenedioxy)bis(ethylamine) has been used to measure biocides released into the environmental water samples.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

249.8 °F - closed cup

Flash Point(C)

121 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Weijie Jiang et al.
ACS applied materials & interfaces, 6(4), 2629-2638 (2014-02-08)
Raspberry-like (RB) polymer particles were prepared, fluorinated, and cast onto glass plates to yield highly water- and oil-repellant superamphiphobic particulate coatings. To procure the RB particles, glycidyl-bearing 212 and 332 nm particles (abbreviated as s-GMA and l-GMA, respectively) were first
Neomycin-Neomycin Dimer: An All-Carbohydrate Scaffold with High Affinity for AT-Rich DNA Duplexes
Kumar S, et al.
Journal of the American Chemical Society, 133(19), 7361-7375 null
Zhangsen Yu et al.
Journal of biomaterials applications, 28(2), 232-240 (2012-04-26)
Fluorescence imaging technique has been used for imaging of biological cells and tissues in vivo. The Cd-free luminescent quantum dots conjugating with a cancer targeting ligand has been taken as a promising biocompatibility and low cytotoxicity system for targeted cancer imaging.
Tamara Buchberger et al.
Journal of chromatography. A, 1318, 22-26 (2013-10-29)
This paper demonstrates the determination of residues of biocidal oligoguanidines manufactured by polycondensation of guanidine hydrochloride and 2,2'-(ethylenedioxy)bis(ethylamine) in environmental water samples. The analytes were preconcentrated from samples adjusted to pH 4 by solid-phase extraction using a mixed-mode sorbent with
Parinaz Fathi et al.
ACS nano, 13(7), 7690-7704 (2019-06-28)
Photoacoustic imaging has emerged as a promising imaging platform with a high tissue penetration depth. However, biodegradable nanoparticles, especially those for photoacoustic imaging, are rare and limited to a few polymeric agents. The development of such nanoparticles holds great promise

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service