Skip to Content
Merck
All Photos(1)

Key Documents

D152609

Sigma-Aldrich

2,2-Dimethylbutyric acid

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5C(CH3)2CO2H
CAS Number:
Molecular Weight:
116.16
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

96%

form

liquid

refractive index

n20/D 1.4154 (lit.)

bp

94-96 °C/5 mmHg (lit.)

density

0.928 g/mL at 25 °C (lit.)

SMILES string

CCC(C)(C)C(O)=O

InChI

1S/C6H12O2/c1-4-6(2,3)5(7)8/h4H2,1-3H3,(H,7,8)

InChI key

VUAXHMVRKOTJKP-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

174.2 °F - closed cup

Flash Point(C)

79 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Abdullah Kutlar et al.
American journal of hematology, 87(11), 1017-1021 (2012-08-14)
Therapeutics which reduce the pathology in sickle cell syndromes are needed, particularly noncytotoxic therapeutics. Fetal hemoglobin (HbF, α(2) γ(2) ) is established as a major regulator of disease severity; increased HbF levels correlate with milder clinical courses and improved survival.
Brandon C Presley et al.
Rapid communications in mass spectrometry : RCM, 34(22), e8908-e8908 (2020-07-28)
Indazole carboxamide synthetic cannabinoids, a prevalent class of recreational drugs, are a major clinical, forensic and public health challenge. One such compound, 5F-ADB, has been implicated in fatalities worldwide. Understanding its metabolism and distribution facilitates the development of laboratory assays
Susan P Perrine et al.
Journal of clinical pharmacology, 51(8), 1186-1194 (2011-03-23)
Pharmacologic induction of fetal globin synthesis is an accepted therapeutic strategy for treatment of the beta hemoglobinopathies and thalassemias, as even small increases in hemoglobin F (HbF) levels reduce clinical severity in sickle cell disease (SCD) and reduce anemia in
Ruoting Xu et al.
Frontiers in immunology, 11, 1994-1994 (2020-09-26)
Encephalitis, the inflammation of the brain, may be caused by an infection or an autoimmune reaction. However, few researches were focused on the gut microbiome characteristics in encephalitis patients. A prospective observational study was conducted in an academic hospital in
Bridgette Wilson et al.
The American journal of gastroenterology, 115(6), 906-915 (2020-05-21)
The low FODMAP diet (LFD) reduces symptoms and bifidobacteria in irritable bowel syndrome (IBS). β-galactooligosaccharides (B-GOS) may reduce the symptoms and increase bifidobacteria in IBS. We investigated whether B-GOS supplementation alongside the LFD improves IBS symptoms while preventing the decline

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service