B88403
3,3-Dimethylbutyric acid
98%
Synonym(s):
tert-Butylacetic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
(CH3)3CCH2COOH
CAS Number:
Molecular Weight:
116.16
Beilstein:
1740427
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
98%
form
liquid
refractive index
n20/D 1.411 (lit.)
bp
185-190 °C (lit.)
mp
−11 °C (lit.)
density
0.912 g/mL at 25 °C (lit.)
SMILES string
CC(C)(C)CC(O)=O
InChI
1S/C6H12O2/c1-6(2,3)4-5(7)8/h4H2,1-3H3,(H,7,8)
InChI key
MLMQPDHYNJCQAO-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
192.2 °F - closed cup
Flash Point(C)
89 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Elham Zahiri et al.
Journal of chromatography. A, 1627, 461390-461390 (2020-08-22)
A dispersive solid phase extraction method was combined with deep eutectic solvent-based solidification of floating organic drop-dispersive liquid-liquid microextraction and used for the extraction/preconcentration of some organophosphorus pesticides residues from edible oil samples. The extracted analytes were quantified with gas
Alex J Krotulski et al.
Drug testing and analysis, 13(2), 427-438 (2020-10-01)
A new class of synthetic cannabinoids has emerged as new psychoactive substances (NPS). Similar in structure to JWH-022, these substances contain alkene modifications to the tail region of the synthetic cannabinoid core structure, and nomenclature denotes these new analogues as
Alex J Krotulski et al.
Journal of forensic sciences, 64(5), 1451-1461 (2019-07-02)
This is the first report regarding the characterization of the new synthetic cannabinoid 4F-MDMB-BINACA. 4F-MDMB-BINACA was first analytically confirmed in seized drug material using gas chromatography-mass spectrometry (GC-MS), liquid chromatography-quadrupole time-of-flight mass spectrometry (LC-QTOF), and nuclear magnetic resonance (NMR) spectroscopy.
James Chun Yip Chan et al.
Journal of pharmaceutical and biomedical analysis, 138, 43-53 (2017-02-09)
A novel liquid chromatography tandem mass spectrometry (LCMSMS) method for the quantitative measurement of gut microbial-derived short-chain fatty acids (SCFAs) in human infant stool has been developed and validated. Baseline chromatographic resolution was achieved for 12 SCFAs (acetic, butyric, caproic
Wen Lie et al.
Archives of toxicology, 95(2), 489-508 (2020-11-26)
5F-MDMB-PINACA and 4F-MDMB-BINACA are synthetic cannabinoids (SCs) that elicit cannabinoid psychoactive effects. Defining pharmacokinetic-pharmacodynamic (PK-PD) relationships governing SCs and their metabolites are paramount to investigating their in vivo toxicological outcomes. However, the disposition kinetics and cannabinoid receptor (CB) activities of
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service