Skip to Content
Merck
All Photos(2)

Key Documents

C5459

Sigma-Aldrich

N-Carbethoxyphthalimide

96%

Synonym(s):

N-Ethoxycarbonylphthalimide, Ethyl 1,3-dioxo-2-isoindolinecarboxylate, Ethyl phthalimide-N-carboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H9NO4
CAS Number:
Molecular Weight:
219.19
Beilstein:
196340
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
crystals
Assay:
96%

Quality Level

Assay

96%

form

crystals

mp

90-92 °C (lit.)

SMILES string

CCOC(=O)N1C(=O)c2ccccc2C1=O

InChI

1S/C11H9NO4/c1-2-16-11(15)12-9(13)7-5-3-4-6-8(7)10(12)14/h3-6H,2H2,1H3

InChI key

VRHAQNTWKSVEEC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Used to protect amine functional groups.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Tetrahedron, 49, 13-13 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service