Skip to Content
Merck
All Photos(1)

Documents

C102814

Sigma-Aldrich

2-Cyclohexen-1-one

≥95%

Synonym(s):

1-Cyclohexen-3-one, 2-Cyclohexenone, 3-Oxocyclohexene, Cyclohexen-3-one, Cyclohexenone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H8(=O)
CAS Number:
Molecular Weight:
96.13
Beilstein:
1280477
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

760 mmHg ( 168 °C)

Assay

≥95%

form

liquid

refractive index

n20/D 1.488 (lit.)

bp

171-173 °C (lit.)

mp

−53 °C (lit.)

density

0.993 g/mL at 25 °C (lit.)

SMILES string

O=C1CCCC=C1

InChI

1S/C6H8O/c7-6-4-2-1-3-5-6/h2,4H,1,3,5H2

InChI key

FWFSEYBSWVRWGL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Versatile electrophile employed in a range of addition reactions including conjugate addition of organocopper nucleophiles, Michael reaction with enol silanes, and phosphoniosilylations.

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 2 Dermal - Acute Tox. 3 Oral - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

132.8 °F - closed cup

Flash Point(C)

56 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron Letters, 34, 3881-3881 (1993)
Albert Poater et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 16(48), 14348-14353 (2010-11-18)
The current approach to improve and tune the enantioselective performances of transition-metal catalysts for asymmetric synthesis is mostly focused to modifications of the steric properties of the ancillary ligands of the active metal. Nevertheless, it is also known that electrostatic
Yan-Hong He et al.
The Journal of organic chemistry, 77(1), 200-207 (2011-11-24)
The direct three-component aza-Diels-Alder reaction of aromatic aldehyde, aromatic amine, and 2-cyclohexen-1-one was catalyzed by hen egg white lysozyme for the first time. Under the optimized conditions investigated in this paper, the enzyme-catalyzed aza-Diels-Alder reaction gave yields up to 98%
Masaki Okamoto et al.
Chemical communications (Cambridge, England), (47)(47), 7363-7365 (2009-12-22)
Conjugate addition of Et(2)Zn to 2-cyclohexen-1-one catalyzed by Cu(OTf)(2) combined with an azolium salt derived from (S)-leucinol produced the corresponding (S)-adduct, while the use of Cu(acac)(2) in combination with the same ligand afforded the (R)-adduct as a major product.
M Teresa Barros et al.
Chemical communications (Cambridge, England), 48(88), 10901-10903 (2012-10-02)
An iodine mediated aromatization leading to a one-pot synthesis of iodo-N-arylanilines and N-arylanilines is reported. This highly regioselective aliphatic-aromatic transformation can be performed with various combinations of 2-cyclohexenones and anilines. The presence of a directing group is crucial for achieving

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service