Skip to Content
Merck
All Photos(1)

Key Documents

712124

Sigma-Aldrich

trans-1-Phenyl-1,3-butadiene

≥95.0% (HPLC)

Synonym(s):

[(E)-1,3-Butadienyl]benzene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H10
CAS Number:
Molecular Weight:
130.19
Beilstein:
1901458
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

≥95.0% (HPLC)

form

liquid

contains

~0.2% 2,6-di-tert-butyl-4-methylphenol as stabilizer

functional group

phenyl

storage temp.

−20°C

SMILES string

C=C\C=C\c1ccccc1

InChI

1S/C10H10/c1-2-3-7-10-8-5-4-6-9-10/h2-9H,1H2/b7-3+

InChI key

XZKRXPZXQLARHH-XVNBXDOJSA-N

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

131.0 °F

Flash Point(C)

55 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service