Skip to Content
Merck
All Photos(2)

Key Documents

B99405

Sigma-Aldrich

2-tert-Butylphenol

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CC6H4OH
CAS Number:
Molecular Weight:
150.22
Beilstein:
1907120
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

0.05 mmHg ( 20 °C)

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.523 (lit.)

bp

224 °C (lit.)

mp

−7 °C (lit.)

density

0.978 g/mL at 25 °C (lit.)

SMILES string

CC(C)(C)c1ccccc1O

InChI

1S/C10H14O/c1-10(2,3)8-6-4-5-7-9(8)11/h4-7,11H,1-3H3

InChI key

WJQOZHYUIDYNHM-UHFFFAOYSA-N

Gene Information

mouse ... Esr1(13982)

Looking for similar products? Visit Product Comparison Guide

Related Categories

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

215.6 °F - Pensky-Martens closed cup

Flash Point(C)

102 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stefano Gabriele et al.
Biomarkers : biochemical indicators of exposure, response, and susceptibility to chemicals, 19(6), 463-470 (2014-07-11)
The aromatic compound p-cresol (4-methylphenol) has been found elevated in the urines of Italian autistic children up to 8 years of age. The present study aims at replicating these initial findings in an ethnically distinct sample and at extending them
Nunna V Krishna et al.
Dalton transactions (Cambridge, England : 2003), 46(3), 770-779 (2016-12-21)
Acid-mediated synthesis of ordered mesoporous aluminosilicates (OMAS) with medium-to-strong Brønsted acid sites and trivalent aluminium exclusively in a tetrahedral framework structure is realized by a newly devised intrinsic hydrolysis method. In this way, we have synthesized a series of well-ordered
Y Yasuda et al.
Microbiology and immunology, 28(11), 1203-1210 (1984-01-01)
Glucose interfered with the inhibitory action of hydrophobic compounds, such as n-octanol, diphenylamine and 2-tert-butylphenol, during L-alanine-initiated germination of Bacillus subtilis spores. The action of glucose on the action of the hydrophobic compounds was not competitive, and the binding affinity
A D Rahimtula et al.
British journal of cancer, 45(6), 935-944 (1982-06-01)
Six phenols [2(3)-t-butyl-4-hydroxyanisole (BHA), 2-t-butylphenol, 4-methoxyphenol, 4-methylmercaptophenol, t-butylhydroquinone and 2,6-di-t-butylphenol] previously shown to be inhibitors of benzo(a)pyrene-induced neoplasia, were examined for their ability to induce in vivo changes in hepatic mono-oxygenase and detoxication enzyme activities, and to act as mono-oxygenase
Zonglin Yang et al.
Journal of hazardous materials, 359, 203-212 (2018-07-24)
Nonylphenols (NPs) are known as Endocrine Disputing Chemicals (ECDs) and Persistent Organic Pollutants (POPs) and have attracted continuous attention. Biodegradation is one of the effective ways for pollutant removal in aquatic, sedimentary and soil environments. In this study, two estuarine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service