Skip to Content
Merck
All Photos(2)

Documents

B67570

Sigma-Aldrich

1-Bromoheptane

99%

Synonym(s):

Heptyl bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)6Br
CAS Number:
Molecular Weight:
179.10
Beilstein:
1697160
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

6.18 (vs air)

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.4499 (lit.)

bp

180 °C (lit.)

mp

−58 °C (lit.)

density

1.14 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCBr

InChI

1S/C7H15Br/c1-2-3-4-5-6-7-8/h2-7H2,1H3

InChI key

LSXKDWGTSHCFPP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

140.0 °F - closed cup

Flash Point(C)

60 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jalal Pourahmad et al.
Iranian journal of pharmaceutical research : IJPR, 11(1), 265-276 (2012-01-01)
Chloroacetaldehyde (CAA) is a chlorination by-product in finished drinking water and a toxic metabolite of a wide variety of industrial chemicals (e.g. vinyl chloride) and chemotherapeutic agents (e.g. cyclophosphamide and ifosfamide). In this research, the cytotoxic mechanisms of CAA in
Beata Gabrić et al.
TheScientificWorldJournal, 2013, 512953-512953 (2013-07-12)
Liquid-liquid extraction is an alternative method that can be used for desulfurization and denitrification of gasoline and diesel fuels. Recent approaches employ different ionic liquids as selective solvents, due to their general immiscibility with gasoline and diesel, negligible vapor pressure
Mehdi Rajabnia Khansari et al.
Environmental science and pollution research international, 24(29), 23382-23388 (2017-08-27)
Perfluorooctanesulfonate (PFOS), an anthropogenic fluorosurfactant, is one of the most common global pollutants. PFOS is used in various consumer products to provide soil, oil, and water resistance to materials used in clothing, upholstery, and food packaging. PFOS is persistent, bioaccumulative
Hypomethylating agents (HMAs) effect on myelodysplastic/myeloproliferative neoplasm unclassifiable (MDS/MPN-U): single institution experience.
Aref Al-Kali et al.
Leukemia & lymphoma, 1-3 (2018-02-22)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service