Skip to Content
Merck
All Photos(1)

Documents

T2265

Sigma-Aldrich

Tetraethylammonium chloride

≥98% (titration)

Synonym(s):

TEA chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)4N(Cl)
CAS Number:
Molecular Weight:
165.70
Beilstein:
3563247
EC Number:
MDL number:
UNSPSC Code:
12352107
PubChem Substance ID:
NACRES:
NA.32

biological source

synthetic (organic)

Assay

≥98% (titration)

form

powder

technique(s)

blocking: suitable

color

white to light yellow

SMILES string

[Cl-].CC[N+](CC)(CC)CC

InChI

1S/C8H20N.ClH/c1-5-9(6-2,7-3)8-4;/h5-8H2,1-4H3;1H/q+1;/p-1

InChI key

YMBCJWGVCUEGHA-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Tetraethylammonium chloride (TEAC) has been used:
  • used for inhibition of peroxynitrite induced relaxation in rat aorta rings
  • as a pharmacological blocker of K+ current and Ca2+ induced K+ current in antennal lobe neurons
  • for the induction of TEAC sensitive currents in cochlear inner hair cells digested with proteolytic enzymes and analyze its properties

Biochem/physiol Actions

Blocks K+ channels; blocks nicotinic acetylcholine neurotransmission by blocking the receptor-mediated K+ currents.
Tetraethylammonium chloride (TEAC) blocks K+ channels non-specifically. In rat aorta rings, tetraethylammonium inhibits relaxation induced by peroxynitrite. It blocks nicotinic acetylcholine neurotransmission by blocking the receptor-mediated K+ currents. TEAC can increase the contractility and mobility of colon and rectum and is a therapeutic option for Hirschsprung′s disease patients. TEAC reduces the inflammatory responses, improves cardiac, vascular and hemodynamic properties during early sepsis in animals. TEAC has cytotoxic and anti-proliferative potential and induces apoptosis in glioma cells by downregulating B-cell lymphoma-2 (Bcl-2) and upregulating Bcl-2 associated X (Bax).

Caution

moisture sensitive, handle under argon

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

201.2 °F - closed cup

Flash Point(C)

94 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Early potassium channel blockade improves sepsis-induced organ damage and cardiovascular dysfunction
Sordi R, et al.
British Journal of Pharmacology, 163(6), 1289-1301 (2011)
Tetraethylammonium enhances the rectal and colonic motility in rats and human in vitro
Li Z, et al.
Naunyn-Schmiedeberg'S Archives of Pharmacology, 384(2), 147-155 (2011)
Dissociation enzyme effects on the potassium currents of inner hair cells isolated from guinea-pig cochlea
Kimitsuki T, et al.
Hearing Research, 199(1-2), 135-139 (2005)
Peroxynitrite-induced relaxation in isolated rat aortic rings and mechanisms of action
Li J, et al.
Toxicology and Applied Pharmacology, 209(3), 269-276 (2005)
Differences of Ca2+ handling properties in identified central olfactory neurons of the antennal lobe
Pippow A, et al.
Cell Calcium, 46(2), 87-98 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service