Skip to Content
Merck
All Photos(2)

Key Documents

A16300

Sigma-Aldrich

N-Acetylglycine

ReagentPlus®, 99%

Synonym(s):

Acetamidoacetic acid, Aceturic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CONHCH2CO2H
CAS Number:
Molecular Weight:
117.10
Beilstein:
774114
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.22

product line

ReagentPlus®

Assay

99%

form

powder or crystals

reaction suitability

reaction type: C-H Activation
reaction type: solution phase peptide synthesis
reagent type: ligand
reaction type: Peptide Synthesis

color

white

mp

207-209 °C (lit.)

application(s)

detection
peptide synthesis

functional group

amine
carboxylic acid

SMILES string

CC(=O)NCC(O)=O

InChI

1S/C4H7NO3/c1-3(6)5-2-4(7)8/h2H2,1H3,(H,5,6)(H,7,8)

InChI key

OKJIRPAQVSHGFK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anatoliy Volkov et al.
Journal of computational chemistry, 25(7), 921-934 (2004-03-18)
Accurate and fast evaluation of electrostatic interactions in molecular systems is still one of the most challenging tasks in the rapidly advancing field of macromolecular chemistry, including molecular recognition, protein modeling and drug design. One of the most convenient and
Jamie L Snow et al.
Journal of the American Chemical Society, 129(32), 9910-9917 (2007-07-26)
Both theoretical and experimental studies are reported for the gas-phase reactions of protonated hydroxylamine with acetic and propanoic acids which yield protonated glycine and alanine, GlyH+ and AlaH+, respectively. The key step for these reactions is an insertion of the
Liangxing Wu et al.
Journal of the American Chemical Society, 130(12), 4089-4096 (2008-03-07)
Eight B-containing compounds, i.e., 1a-h, were prepared as mimics of the green fluorescent protein (GFP) fluorophore. The underlying concept was that synthetic GFP chromophore analogues are not fluorescent primarily because of free rotation about an aryl-alkene bond (Figure 1b). This
S K Mehta et al.
Journal of colloid and interface science, 314(2), 689-698 (2007-06-26)
The well-characterized, monodisperse nature of reverse micelles formed by sodium bis-(2-ethylhexyl)sulfosuccinate/water/isooctane and their usefulness in assimilating compounds of varied interests have been exploited to investigate the effect of acetyl modified amino acids (MAA) viz., N-acetyl-L-glycine (NAG), N-acetyl-L-aspartic acid (NAA) and
H K Ogawa et al.
Journal of chromatography, 612(1), 145-149 (1993-01-29)
A simple and sensitive method for the determination of N-acetylneuraminic acid and N-glycolylneuraminic acid in glycoproteins by high-performance liquid chromatography has been investigated. The best condition to release sialic acid from glycoproteins involved incubation at 80 degrees C for 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service