Skip to Content
Merck
All Photos(1)

Key Documents

928356

Sigma-Aldrich

XantPhos ChemBeads

Synonym(s):

4,5-Bis(diphenylphosphino)-9,9-dimethylxanthene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C39H32OP2
CAS Number:
Molecular Weight:
578.62
MDL number:
UNSPSC Code:
12352005
NACRES:
NA.82
Pricing and availability is not currently available.

form

solid

Quality Level

composition

~ 4 wt.% loading of catalyst

reaction suitability

reagent type: catalyst
reaction type: Cross Couplings

SMILES string

CC1(c2c(Oc3c1cccc3P(c4ccccc4)c5ccccc5)c(P(c6ccccc6)c7ccccc7)ccc2)C

InChI

1S/C39H32OP2/c1-39(2)33-25-15-27-35(41(29-17-7-3-8-18-29)30-19-9-4-10-20-30)37(33)40-38-34(39)26-16-28-36(38)42(31-21-11-5-12-22-31)32-23-13-6-14-24-32/h3-28H,1-2H3

InChI key

CXNIUSPIQKWYAI-UHFFFAOYSA-N

Application

  • Ligand used in a synthesis of heterocycles by palladium-catalyzed C-N cross coupling of 3-bromothiophenes with 2-aminopyridines. Also used in a ruthenium-catalyzed alkylation of active methylene compounds with alcohols.
  • Metal chelating ligand for catalysis.
  • ChemBeads are chemical coated glass beads. ChemBeads offer improved flowability and chemical uniformity perfect for automated solid dispensing and high-throughput experimentation. The method of creating ChemBeads uses no other chemicals or surfactants allowing the user to accurately dispense sub-milligram amounts of chemical.
Learn more about ChemBeads products

For larger scale uses, product also available in powdered form (526460)

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B Inhalation

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service