927791
Bis(di-tert-butyl(4-dimethylaminophenyl)phosphine)dichloropalladium(II) ChemBeads
Synonym(s):
(A-taPhos)2PdCl2, Pd(amphos)Cl2
About This Item
form
solid
Quality Level
composition
~ 5 wt.% loading of catalyst
reaction suitability
reagent type: catalyst
reaction type: Cross Couplings
SMILES string
Cl[Pd]Cl.CN(C)c1ccc(cc1)P(C(C)(C)C)C(C)(C)C.CN(C)c2ccc(cc2)P(C(C)(C)C)C(C)(C)C
InChI
1S/2C16H28NP.2ClH.Pd/c2*1-15(2,3)18(16(4,5)6)14-11-9-13(10-12-14)17(7)8;;;/h2*9-12H,1-8H3;2*1H;/q;;;;+2/p-2
InChI key
DWOZNANUEDYIOF-UHFFFAOYSA-L
Application
Palladium-catalyzed coupling of iodo-α-β-unsaturated esters to afford tetrasubstituted olefins
ChemBeads are chemical coated glass beads. ChemBeads offer improved flowability and chemical uniformity perfect for automated solid dispensing and high-throughput experimentation. The method of creating ChemBeads uses no other chemicals or surfactants allowing the user to accurately dispense sub-milligram amounts of chemical.
Learn more about ChemBeads products
For larger scale uses, product also available in powdered form (678740)
Other Notes
Versatile Methods to Dispense Sub-Milligram Quantities of Solids using Chemical Coated Beads for High-Throughput Experimentation
ChemBead Enabled High-Throughput Cross-Electrophile Coupling Reveals a New Complementary Ligand
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Carc. 1B Inhalation
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service