Skip to Content
Merck
All Photos(3)

Documents

919764

Sigma-Aldrich

Grubbs Catalyst® M204 ChemBeads

Synonym(s):

(1,3-Bis(2,4,6-trimethylphenyl)-2-imidazolidinylidene)dichloro(phenylmethylene)(tricyclohexylphosphine)ruthenium, Benzylidene[1,3-bis(2,4,6-trimethylphenyl)-2-imidazolidinylidene]dichloro(tricyclohexylphosphine)ruthenium, Dichloro[1,3-bis(2,4,6-trimethylphenyl)-2-imidazolidinylidene](benzylidene)(tricyclohexylphosphine)ruthenium(II), Grubbs Catalyst® 2nd Generation, Grubbs Catalyst® M2a (C848)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C46H65Cl2N2PRu
CAS Number:
Molecular Weight:
848.97
UNSPSC Code:
12352103
NACRES:
NA.22

form

solid

Quality Level

composition

~ 4 wt.% loading of catalyst

reaction suitability

reagent type: catalyst
core: ruthenium
reaction type: Ring-Opening Polymerization

storage temp.

2-8°C

SMILES string

CC1=CC(C)=CC(C)=C1N(CCN2C3=C(C=C(C=C3C)C)C)[C-2]2=[Ru+6](P(C4CCCCC4)(C5CCCCC5)C6CCCCC6)([Cl-])([Cl-])=[C-2]C7=CC=CC=C7

InChI

1S/C21H26N2.C18H33P.C7H6.2ClH.Ru/c1-14-9-16(3)20(17(4)10-14)22-7-8-23(13-22)21-18(5)11-15(2)12-19(21)6;1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18;1-7-5-3-2-4-6-7;;;/h9-12H,7-8H2,1-6H3;16-18H,1-15H2;1-6H;2*1H;/q;;;;;+2/p-2

InChI key

FCDPQMAOJARMTG-UHFFFAOYSA-L

Application

Grubbs Catalyst® M204 can be used as a catalyst for ring-closing metathesis (RCM), cross-metathesis, and ring-opening metathesis polymerization (ROMP). It is also used to synthesize trisubstituted olefins with excellent functional group tolerance and selectivity via cross-metathesis and ring closing metathesis reactions.
It can also be used as a catalyst:
To synthesize coumarins from phenolic compounds via RCM.
To cleave secondary (E)-allyl vic-diols to aldehydes.

ChemBeads are chemical coated glass beads. ChemBeads offer improved flowability and chemical uniformity perfect for automated solid dispensing and high-throughput experimentation. The method of creating ChemBeads uses no other chemicals or surfactants allowing the user to accurately dispense sub-milligram amounts of chemical.
Learn more about ChemBeads products

For larger scale uses, product is also available in powdered form (569747)

Legal Information

Grubbs Catalyst is a registered trademark of Umicore AG & Co. KG

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B Inhalation - Flam. Sol. 2

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Versatile Methods to Dispense Sub-Milligram Quantities of Solids using Chemical Coated Beads for High-Throughput Experimentation.
Martin CM, et al.
Organic Process Research & Development, 23(9), 1900-1907 (2019)
Highly selective cross-metathesis with phenyl vinyl sulphone using the ′second generation′Grubbs′ catalyst
Grela K and Bieniek M
Tetrahedron Letters, 42(36), 6425-6428 (2001)
Synthesis of coumarins by ring-closing metathesis using Grubbs' catalyst
Van Tuyen N, et al.
Tetrahedron Letters, 44(22), 4199-4201 (2003)
Unusual influence of substituents on ring-opening metathesis reactions
Wright D L, et al.
Organic Letters, 3(26), 4275-4277 (2002)
Reversible inhibition/activation of olefin metathesis: a kinetic investigation of ROMP and RCM reactions with Grubbs' catalyst
P'Poo SJ and Schanz H-J
Journal of the American Chemical Society, 129(46), 14200-14212 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service