Skip to Content
Merck
All Photos(1)

Key Documents

92749

Sigma-Aldrich

2-(Trimethylsilyl)ethoxymethyl chloride

≥95.0% (GC)

Synonym(s):

(2-Chloromethoxyethyl)trimethylsilane, Chloromethyl 2-trimethylsilylethyl ether, SEM-Cl, SEM-chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3SiCH2CH2OCH2Cl
CAS Number:
Molecular Weight:
166.72
Beilstein:
3587289
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95.0% (GC)

form

liquid

refractive index

n20/D 1.435 (lit.)
n20/D 1.436

bp

170-172 °C (lit.)
57-59 °C/8 mmHg (lit.)

density

0.942 g/mL at 25 °C (lit.)

functional group

chloro
ether

shipped in

wet ice

storage temp.

−20°C

SMILES string

C[Si](C)(C)CCOCCl

InChI

1S/C6H15ClOSi/c1-9(2,3)5-4-8-6-7/h4-6H2,1-3H3

InChI key

BPXKZEMBEZGUAH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-(Trimethylsilyl)ethoxymethyl chloride (SEM-Cl) is a widely used reagent for the preparation protection groups for amines, alcohols, phenols, and carboxy groups. The nitrogen of amides and sulfonamides groups are also protected by using SEM-Cl reagent.

Application

SEM-Cl can be used as a reagent for the preparation of 2,4,5-trisubstituted tetrahydropyrans form homoallylic alcohols by a Prins-type cyclization reaction. It can also be used in the synthesis of amino acid SEM esters from N-protected amino acids in the presence of lithium carbonate.

Other Notes

Protecting-group reagent for alcohols and other functional groups selectively removable with Bu4NF ; Protecting group for pyrroles and imidazoles, facilitating selective metalation ; Reagent for the introduction of a protected C1 building block

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

114.8 °F - closed cup

Flash Point(C)

46 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T. Wada et al.
Tetrahedron Letters, 36, 1683-1683 (1995)
Protecting groups for the pyrrole and indole nitrogen atom. The [2-(trimethylsilyl) ethoxy] methyl moiety. Lithiation of 1-[[2-(trimethylsilyl) ethoxy] methyl] pyrrole.
Muchowski J, et al.
The Journal of Organic Chemistry, 49(1), 203-205 (1984)
K Kim et al.
Chemical communications (Cambridge, England), (21), 2268-2269 (2002-09-21)
A synthetic method to prepare a novel polymer-supported 2-(diphenylmethylsilyl)ethoxymethyl chloride (DSEM-Cl) linker and its applications are described.
B.H. Lipshutz et al.
Tetrahedron Letters, 30, 7149-7149 (1989)
D. Crich et al.
Synlett, 117-117 (1990)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service