Skip to Content
Merck
All Photos(1)

Key Documents

100331

Sigma-Aldrich

Chloromethyl methyl ether

technical grade

Synonym(s):

MOM chloride, Methoxymethyl chloride, Methyl chloromethyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2OCH3
CAS Number:
Molecular Weight:
80.51
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

vapor pressure

3.55 psi ( 20 °C)

Assay

≥92.5%

form

liquid

refractive index

n20/D 1.396 (lit.)

bp

55-57 °C ((lit.))
55-57 °C (lit.)

density

1.06 g/mL at 25 °C (lit.)
1.06 g/mL at 25 °C

functional group

chloro
ether

storage temp.

2-8°C

SMILES string

COCCl

InChI

[1S/C2H5ClO/c1-4-2-3/h2H2,1H3]
1S/C2H5ClO/c1-4-2-3/h2H2,1H3

InChI key

[XJUZRXYOEPSWMB-UHFFFAOYSA-N]
XJUZRXYOEPSWMB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Chloromethyl methyl ether (MOM-Cl) can be used as a reagent:
  •  For the introduction of the methoxymethyl (MOM) protecting group by converting alcohols to methoxy methyl (MOM) ethers.
  •  To prepare some methoxymethyl group containing metal complexes coordinated to carbonyl and dipyridyl ligands.        
  • In the preparation of fluorene/chloromethyl methyl ether cross-linked polymers by Friedel-Crafts polymerization.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Carc. 1A - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

60.8 °F - closed cup

Flash Point(C)

16 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jung-Jae Lee et al.
Chemical communications (Cambridge, England), (15)(15), 1962-1963 (2009-04-01)
N-Alkylation of a fluorescent macrocyclic amine in aqueous micellar solution produces enhanced emission; the reaction with chloromethyl methyl ether exhibits autocatalysis.
Bis(Chloromethyl) ether and technical-grade chloromethyl methyl ether.
Report on carcinogens : carcinogen profiles, 11, III56-III57 (2004-01-01)
A Blair et al.
Cancer causes & control : CCC, 8(3), 473-490 (1997-05-01)
Epidemiologic evidence on the relation between reactive chemicals and cancer is reviewed. These highly reactive chemicals (acrylonitrile; bis[chloromethyl]ether and chloromethyl methyl ether; 1,3-butadiene, ethylene oxide; formaldehyde; mustard gas; sulfuric acid; and vinyl chloride) vary in use and exposure. All are
A modified low-cost preparation of chloromethyl methyl ether (MOM-Cl)
Reggelin M and Doerr S
Synlett, 2004(06), 1117-1117 (2004)
Organometallics, 25, 4909-4909 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service