Skip to Content
Merck
All Photos(3)

Key Documents

926485

Sigma-Aldrich

Phase Guide Mixer Chip

Fluidic 533, COC

Synonym(s):

Microfluidic chip

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
42142600
NACRES:
NA.23

description

Microfludic chip x1

Application

The Phase Guide Mixer Fluidic 533 COC microfluidic chip applies passive phaseguided mixing principles and is not meant to be used in flow-through mode.

Features and Benefits

  • Three varying micromixing units per chip
  • Each unit features three inlets and one outlet
  • Phaseguide technology provides complete control over the filling and emptying of microfluidic structures

Components

Chip Properties:
  • Mini Luer Interface
  • Material: Cyclic olefin copolymer (COC)
  • Volume of Chambers A & B: 5.6 μl
  • Volume of Chambers C & D: 13.2 μl
  • Volume of Chambers E & F: 11.1 μl

This mixer is based on the principles described in S. Hakenberg et al., A phaseguided passive batch microfluidic mixing chamber for isothermal amplification, Lab Chip, 12, 4576-4580, 2012.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dongfei Liu et al.
Lab on a chip, 17(11), 1856-1883 (2017-05-10)
The microfluidic technique has brought unique opportunities toward the full control over the production processes for drug delivery carriers, owing to the miniaturisation of the fluidic environment. In comparison to the conventional batch methods, the microfluidic setup provides a range
Sharma T Sanjay et al.
Advanced drug delivery reviews, 128, 3-28 (2017-09-19)
Conventional systematically-administered drugs distribute evenly throughout the body, get degraded and excreted rapidly while crossing many biological barriers, leaving minimum amounts of the drugs at pathological sites. Controlled drug delivery aims to deliver drugs to the target sites at desired

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service