Skip to Content
Merck
All Photos(3)

Key Documents

763705

Sigma-Aldrich

Graphene oxide dispersion

greener alternative

2 mg/mL,dispersion in H2O, avg. no. of layers, 1

Synonym(s):

graphene oxide aqueous dispersion

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CxOyHz
UNSPSC Code:
12352103
NACRES:
NA.23

Product Name

Graphene oxide, 2 mg/mL, dispersion in H2O, avg. no. of layers, 1

form

dispersion in H2O

Quality Level

feature

avg. no. of layers 1

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

concentration

2 mg/mL

refractive index

n20/D 1.333

density

0.981 g/mL at 25 °C

greener alternative category

SMILES string

O=C(O)C1C2=C3C4=C5C6=C7C8=C9C%10=C%11C(C%12=C%13C%10=C%14C8=C%15C6=C%16C4=C%17C2=CC(C(O)=O)C%18=C%17C%19=C%16C%20=C%15C%21=C%14C%22=C%13C(C%23=C%24C%22=C%25C%21=C%26C%20=C%27C%19=C%28C%18=CC(C(O)=O)C%29=C%28C%30=C%27C%31=C%26C%32=C%25C%33=C%24C(C%34=C%35C

InChI

1S/C140H42O20/c141-131(142)26-13-23-15-44-62(140(159)160)45-16-24-14-40-31(132(143)144)5-1-29-41-20-48(135(149)150)56-33-7-3-28-27-2-6-32-55-37(133(145)146)11-9-35-60(138(155)156)42-17-25-18-43-61(139(157)158)36-10-12-38(134(147)148)58-46-21-50(137(153)154)59-47-22-49(136(151)152)57-34-8-4-30-39(19-26)51(23)78-72(44)88-75(45)80-52(24)79(54(29)40)95-71(41)83(56)101-93-69(33)64(28)91-90-63(27)68(32)92-86(66(35)55)73(42)81-53(25)82-74(43)87(67(36)58)96-76(46)85(59)103-97-77(47)84(57)102-94-70(34)65(30)89(78)105-104(88)115-98(80)111(95)116(101)126-122-110(93)107(91)120-119-106(90)108(92)99(81)114-100(82)112(96)118(103)128(124(114)119)123-113(97)117(102)127(130(122)129(120)123)121(109(94)105)125(115)126/h2,5,7-10,12-22,26,38,48-50H,1,3-4,6,11H2,(H,141,142)(H,143,144)(H,145,146)(H,147,148)(H,149,150)(H,151,152)(H,153,154)(H,155,156)(H,157,158)(H,159,160)

InChI key

VTWITIAIMADGRM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

  • Chloride free (purified by dialysis)
  • Monolayer sheet
  • sheet diameter <10 micron
Graphene oxide (GO) is a one atom thick 2D carbon material with excellent thermal, mechanical, and electrical properties due to its unique structural and morphological features. GO can be easily dispersed in aqueous and polar organic solvents to facilitate its practical use. It can be synthesized by modified Hummer’s method. It can be applied in the fields of drug delivery, energy storage, oil-water separation, immobilization catalysis, desalination, and solar cells.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product belongs to Enabling category of greener alternatives thus aligns with "Design for energy efficency". High concentrated graphene oxide sheets provide the prerequisite viscosity to bind the electrode materials together and enable 3D printing. Using water as a green solvent makes this aqueous ink system feasible for processing and drying safety and low cost. Click here for more information.

Application

Because of the presence of oxygen-containing functional groups and acidic nature, GO can serve as both carbocatalyst as well as a supporting scaffold for various catalytically active species. GO can facilitate many organic transformations such as the oxidation of sulfides, olefins, and various hydrocarbons, Friedel–Crafts reaction, Aza–Michael additions, condensation, and ring-opening polymerization.

It can be used as a support for biocatalysis in organic solvents. For example, carboxyl-functionalized graphene oxide can act as a support to immobilize Yarrowia lipolytica lipase. The immobilized enzyme exhibits a high efficiency for the resolution of the racemic compound in the organic solvent.

It can also be used as a hole transport layer in organic photovoltaic cells.

Features and Benefits

  • Good solution processability
  • Hydrophilic and easily dispersed in water
  • Low production cost
  • Presence of rich active oxygen-containing functional groups
  • It can be easily functionalized
Large surface area, high chemical stability, good charge carrier properties.

Preparation Note

Product may aggregate over time to form larger particles. Sonicate before use if single layer graphene oxide required.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jin Ok Hwang et al.
ACS nano, 6(1), 159-167 (2011-12-14)
Graphene is a promising candidate to complement brittle and expensive transparent conducting oxides. Nevertheless, previous research efforts have paid little attention to reduced graphene, which can be of great benefit due to low-cost solution processing without substrate transfer. Here we
Michał S Barski et al.
Nature communications, 11(1), 5043-5043 (2020-10-09)
Human T-cell lymphotropic virus type 1 (HTLV-1) is a deltaretrovirus and the most oncogenic pathogen. Many of the ~20 million HTLV-1 infected people will develop severe leukaemia or an ALS-like motor disease, unless a therapy becomes available. A key step
Nitish Sathyanarayanan et al.
Nature communications, 10(1), 4127-4127 (2019-09-13)
Substrate channeling is a mechanism for the internal transfer of hydrophobic, unstable or toxic intermediates from the active site of one enzyme to another. Such transfer has previously been described to be mediated by a hydrophobic tunnel, the use of
N-doped graphitic self-encapsulation for high performance silicon anodes in lithium-ion batteries.
JunaLee, Won, et al.
Energy & Environmental Science, 7(2), 621-626 (2014)
Goki Eda et al.
Advanced materials (Deerfield Beach, Fla.), 22(22), 2392-2415 (2010-05-01)
Chemically derived graphene oxide (GO) possesses a unique set of properties arising from oxygen functional groups that are introduced during chemical exfoliation of graphite. Large-area thin-film deposition of GO, enabled by its solubility in a variety of solvents, offers a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service