Skip to Content
Merck
All Photos(1)

Documents

919527

Sigma-Aldrich

Selenium nanoparticles

150 nm particle size, 0.15 wt. % in water, dispersion

Synonym(s):

SeNP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Se
CAS Number:
Molecular Weight:
78.96
UNSPSC Code:
12141910
NACRES:
NA.23

form

dispersion

Quality Level

concentration

0.15 wt. % in water

particle size

150 nm

InChI

1S/Se

InChI key

BUGBHKTXTAQXES-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Selenium nanoparticles (SeNPs) are used in a wide variety of applications such as antimicrobial coatings, nanotherapeutics, diagnostics, and medical devices. These nanoparticles have been shown to play an important role in protecting against oxidative stress and have demonstrated low toxicity and bioavailability.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Selenium Nanoparticles: Biomedical Applications
Vrcek V I, et al.
Springer Handbook of Nanotechnology (2018)
Amit Khurana et al.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie, 111, 802-812 (2019-01-08)
Nanoparticles (NPs) serve to reduce the toxicity, enhance bioactivity, improve targeting, and provide versatile means to control the release profile of the encapsulated moiety. Among different NPs, inorganic NPs of metals like Ag, Au, Ce, Fe, Se, Ti and Zn

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service