Skip to Content
Merck
All Photos(1)

Documents

8.08352

Sigma-Aldrich

Triethylamine

for synthesis

Synonym(s):

Triethylamine, N,N-Diethylethanamine, TEA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)3N
CAS Number:
Molecular Weight:
101.19
MDL number:
UNSPSC Code:
12352116
EC Index Number:
204-469-4
NACRES:
NA.22

vapor pressure

72 hPa ( 20 °C)

Quality Level

Assay

≥99% (GC)

form

liquid

autoignition temp.

215 °C

potency

730 mg/kg LD50, oral (Rat)

expl. lim.

1.2-9.3 % (v/v)

pH

12.7 (15 °C, 100 g/L in H2O, IUCLID)

bp

90 °C/1013 hPa

mp

-115 °C

transition temp

flash point -11 °C

solubility

133 g/L

density

0.73 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C6H15N/c1-4-7(5-2)6-3/h4-6H2,1-3H3

InChI key

ZMANZCXQSJIPKH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Triethylamine is a volatile organic compound used in several industrial processes such as oxidation, reduction, elimination, substitution, addition, and dehydrohalogenation reactions.

Application

Triethylamine can be used as:
  • A catalyst in the Domino ring-opening/recyclization reactions of 1,3-thiazolidinedione, malononitrile, and aromatic aldehydes to synthesize polysubstituted dihydrothiophenes.
  • A base to synthesize sulfonamides using primary amines and sulfonylating agents.
  • A solvent in the Pd-catalyzed cascade Suzuki-Heck reaction.

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.726 - 0.728
Water (K. F.): ≤ 0.20 %
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

12.2 °F - closed cup

Flash Point(C)

-11 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service