Skip to Content
Merck
All Photos(2)

Key Documents

900891

Sigma-Aldrich

Azo biotin-azide

Synonym(s):

Diazo biotin-azide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H45N9O7S
CAS Number:
Molecular Weight:
711.83
MDL number:
UNSPSC Code:
12352125
NACRES:
NA.22

form

powder or crystals

Quality Level

reaction suitability

reaction type: click chemistry

storage temp.

−20°C

SMILES string

S1[C@H]([C@H]4NC(=O)N[C@H]4C1)CCCCC(=O)NCCOCCOCCOCCNC(=O)c2ccc(cc2)\N=N\c3c(ccc(c3)CCN=[N+]=[N-])O

InChI

1S/C33H45N9O7S/c34-42-37-12-11-23-5-10-28(43)26(21-23)41-40-25-8-6-24(7-9-25)32(45)36-14-16-48-18-20-49-19-17-47-15-13-35-30(44)4-2-1-3-29-31-27(22-50-29)38-33(46)39-31/h5-10,21,27,29,31,43H,1-4,11-20,22H2,(H,35,44)(H,36,45)(H2,38,39,46)/b41-40+/t27-,29-,

InChI key

VQJUOWXTLWKWGJ-GKVYSXGOSA-N

Application

Azo biotin-azide is a linker that contains a biotin moiety connected to an azide group through a spacer arm containing a diazo group that can be cleaved with 50 mM sodium dithionite solution. The biotin can be captured by streptavidin and the azide can react with terminal or strained alkynes.

Automate your Biotin tagging with Synple Automated Synthesis Platform (SYNPLE-SC002)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A chemoselective cleavable fluorescence turn-ON linker for proteomic studies.
Liew S S, et al.
Chemical Communications (Cambridge, England), 53(100), 13332-13335 (2017)
Chemical Modification and Organelle?Specific Localization of Orlistat?Like Natural?Product?Based Probes.
Yang P Y, et al.
Chemistry - An Asian Journal, 6(10), 2762-2775 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service